Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 31289490-31289725 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PIK3IP1
Entrez Gene ID
113791
Gene Name
phosphoinositide-3-kinase interacting protein 1
Gene Aliases
HGFL, hHGFL(S)
UniGene
Hs.26670
Gene Chromosome Location
Chr.22: 31281593-31292534 on Build GRCh38
Transcripts
NM_001135911.1 NM_052880.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1291240 c.326T>C p.L109P Chr.22: 31289681-31289681 on Build GRCh38
1033435 c.343G>T p.E115* Chr.22: 31289664-31289664 on Build GRCh38
1033436 c.331G>A p.A111T Chr.22: 31289676-31289676 on Build GRCh38
5583938 c.321G>A p.Q107Q Chr.22: 31289686-31289686 on Build GRCh38
5482611 c.413C>G p.P138R Chr.22: 31289594-31289594 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs778992121 Chr.22: 31289593-31289593 on Build GRCh38
rs773370421 Chr.22: 31289669-31289669 on Build GRCh38
rs753886272 Chr.22: 31289555-31289555 on Build GRCh38
rs748174538 Chr.22: 31289597-31289597 on Build GRCh38
rs759662375 Chr.22: 31289566-31289566 on Build GRCh38
rs202122937 Chr.22: 31289650-31289650 on Build GRCh38
rs55940931 Chr.22: 31289530-31289530 on Build GRCh38
rs1005544852 Chr.22: 31289571-31289571 on Build GRCh38
rs1054858335 Chr.22: 31289614-31289614 on Build GRCh38
rs553352319 Chr.22: 31289524-31289524 on Build GRCh38
rs893177052 Chr.22: 31289626-31289626 on Build GRCh38
rs866587054 Chr.22: 31289544-31289544 on Build GRCh38
rs368477421 Chr.22: 31289573-31289573 on Build GRCh38
rs372513285 Chr.22: 31289511-31289511 on Build GRCh38
rs752850127 Chr.22: 31289579-31289579 on Build GRCh38
rs775344496 Chr.22: 31289519-31289519 on Build GRCh38
rs769551604 Chr.22: 31289516-31289516 on Build GRCh38
rs981262068 Chr.22: 31289542-31289542 on Build GRCh38
rs765432241 Chr.22: 31289569-31289569 on Build GRCh38
rs148357191 Chr.22: 31289668-31289668 on Build GRCh38
rs751400324 Chr.22: 31289608-31289608 on Build GRCh38
rs761041026 Chr.22: 31289551-31289551 on Build GRCh38
rs376084730 Chr.22: 31289651-31289651 on Build GRCh38
rs868430081 Chr.22: 31289595-31289595 on Build GRCh38
rs773642449 Chr.22: 31289538-31289538 on Build GRCh38

View Full Product Details