Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 98416678-98416931 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RAP1GDS1
Entrez Gene ID
5910
Gene Name
Rap1 GTPase-GDP dissociation stimulator 1
Gene Aliases
GDS1, SmgGDS
UniGene
Hs.132858
Gene Chromosome Location
Chr.4: 98261376-98443861 on Build GRCh38
Transcripts
NM_001100426.1 NM_001100427.1 NM_001100428.1 NM_001100429.1 NM_001100430.1 NM_021159.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4420061 c.767-9G>A p.? Chr.4: 98416736-98416736 on Build GRCh38
6584433 c.826C>G p.Q276E Chr.4: 98416804-98416804 on Build GRCh38
5431143 c.767-8T>C p.? Chr.4: 98416737-98416737 on Build GRCh38
5866983 c.865G>T p.E289* Chr.4: 98416843-98416843 on Build GRCh38
3747603 c.866A>G p.E289G Chr.4: 98416844-98416844 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs201519891 Chr.4: 98416843-98416843 on Build GRCh38
rs758589911 Chr.4: 98416786-98416786 on Build GRCh38
rs142041818 Chr.4: 98416744-98416744 on Build GRCh38
rs1002404243 Chr.4: 98416834-98416834 on Build GRCh38
rs768128355 Chr.4: 98416719-98416719 on Build GRCh38
rs997065535 Chr.4: 98416791-98416791 on Build GRCh38
rs779726252 Chr.4: 98416896-98416896 on Build GRCh38
rs752944807 Chr.4: 98416777-98416777 on Build GRCh38
rs776791878 Chr.4: 98416739-98416739 on Build GRCh38
rs924952913 Chr.4: 98416701-98416701 on Build GRCh38
rs745708668 Chr.4: 98416860-98416860 on Build GRCh38
rs764227821 Chr.4: 98416805-98416805 on Build GRCh38
rs767597464 Chr.4: 98416713-98416713 on Build GRCh38
rs62323576 Chr.4: 98416770-98416770 on Build GRCh38
rs201021594 Chr.4: 98416782-98416782 on Build GRCh38
rs1038493134 Chr.4: 98416764-98416764 on Build GRCh38
rs1013799512 Chr.4: 98416903-98416903 on Build GRCh38
rs574881601 Chr.4: 98416712-98416712 on Build GRCh38
rs891262608 Chr.4: 98416889-98416889 on Build GRCh38
rs781186324 Chr.4: 98416846-98416846 on Build GRCh38
rs769401329 Chr.4: 98416706-98416706 on Build GRCh38
rs765608907 Chr.4: 98416765-98416765 on Build GRCh38
rs10516430 Chr.4: 98416730-98416730 on Build GRCh38
rs775111382 Chr.4: 98416711-98416711 on Build GRCh38
rs745318115 Chr.4: 98416733-98416735 on Build GRCh38
rs771726814 Chr.4: 98416777-98416778 on Build GRCh38
rs371957742 Chr.4: 98416733-98416733 on Build GRCh38
rs759936871 Chr.4: 98416748-98416748 on Build GRCh38
rs940009912 Chr.4: 98416736-98416736 on Build GRCh38
rs751563026 Chr.4: 98416841-98416841 on Build GRCh38
rs761041002 Chr.4: 98416731-98416731 on Build GRCh38
rs369611053 Chr.4: 98416866-98416866 on Build GRCh38
rs113035112 Chr.4: 98416907-98416907 on Build GRCh38

View Full Product Details