Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 104931952-104932183 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PLD4
Entrez Gene ID
122618
Gene Name
phospholipase D family member 4
Gene Aliases
C14orf175
UniGene
Hs.407101
Gene Chromosome Location
Chr.14: 104924806-104937789 on Build GRCh38
Transcripts
NM_001308174.1 NM_138790.3 XM_011536411.2 XM_017020965.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5821720 c.1008-1G>A p.? Chr.14: 104932011-104932011 on Build GRCh38
3885710 c.1186G>A p.A396T Chr.14: 104932139-104932139 on Build GRCh38
6343337 c.1218G>T p.V406V Chr.14: 104932171-104932171 on Build GRCh38
5718728 c.1037G>A p.R346Q Chr.14: 104932041-104932041 on Build GRCh38
697864 c.1042G>T p.A348S Chr.14: 104932046-104932046 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs754321469 Chr.14: 104932011-104932011 on Build GRCh38
rs770428272 Chr.14: 104932041-104932041 on Build GRCh38
rs371939654 Chr.14: 104932111-104932111 on Build GRCh38
rs759628235 Chr.14: 104932157-104932157 on Build GRCh38
rs771834884 Chr.14: 104932032-104932032 on Build GRCh38
rs577930915 Chr.14: 104932060-104932060 on Build GRCh38
rs373382635 Chr.14: 104932151-104932151 on Build GRCh38
rs759416582 Chr.14: 104931976-104931976 on Build GRCh38
rs755486070 Chr.14: 104932016-104932016 on Build GRCh38
rs780421637 Chr.14: 104932098-104932103 on Build GRCh38
rs557134091 Chr.14: 104932153-104932153 on Build GRCh38
rs980452281 Chr.14: 104932124-104932124 on Build GRCh38
rs190449169 Chr.14: 104932137-104932137 on Build GRCh38
rs758765691 Chr.14: 104932084-104932084 on Build GRCh38
rs769308299 Chr.14: 104932117-104932117 on Build GRCh38
rs767786871 Chr.14: 104931993-104931993 on Build GRCh38
rs373643392 Chr.14: 104932070-104932070 on Build GRCh38
rs74316182 Chr.14: 104932055-104932055 on Build GRCh38
rs766460978 Chr.14: 104932068-104932068 on Build GRCh38
rs745428855 Chr.14: 104932114-104932114 on Build GRCh38
rs746674903 Chr.14: 104932035-104932035 on Build GRCh38
rs750901959 Chr.14: 104932061-104932062 on Build GRCh38
rs1031258522 Chr.14: 104932074-104932074 on Build GRCh38
rs775340893 Chr.14: 104932118-104932118 on Build GRCh38
rs779617815 Chr.14: 104932073-104932073 on Build GRCh38
rs899838595 Chr.14: 104932017-104932017 on Build GRCh38
rs778308503 Chr.14: 104932098-104932098 on Build GRCh38
rs1011188055 Chr.14: 104932043-104932043 on Build GRCh38
rs750617640 Chr.14: 104932064-104932064 on Build GRCh38
rs777506608 Chr.14: 104932033-104932033 on Build GRCh38
rs755502080 Chr.14: 104932071-104932071 on Build GRCh38
rs1011069335 Chr.14: 104932099-104932099 on Build GRCh38
rs780026398 Chr.14: 104932005-104932005 on Build GRCh38
rs763279228 Chr.14: 104931975-104931975 on Build GRCh38
rs867343562 Chr.14: 104932148-104932148 on Build GRCh38
rs763883111 Chr.14: 104932063-104932063 on Build GRCh38
rs759489682 Chr.14: 104932054-104932054 on Build GRCh38
rs749252923 Chr.14: 104932119-104932119 on Build GRCh38
rs34261826 Chr.14: 104932050-104932050 on Build GRCh38
rs762019106 Chr.14: 104931992-104931992 on Build GRCh38
rs750659366 Chr.14: 104932003-104932003 on Build GRCh38
rs765063111 Chr.14: 104931984-104931984 on Build GRCh38
rs770533954 Chr.14: 104932110-104932110 on Build GRCh38
rs760827962 Chr.14: 104932067-104932067 on Build GRCh38
rs760918977 Chr.14: 104932146-104932146 on Build GRCh38
rs116640235 Chr.14: 104931974-104931974 on Build GRCh38
rs776057514 Chr.14: 104932044-104932044 on Build GRCh38
rs765409152 Chr.14: 104932159-104932159 on Build GRCh38
rs929333550 Chr.14: 104932136-104932136 on Build GRCh38
rs756150793 Chr.14: 104932004-104932004 on Build GRCh38
rs956442773 Chr.14: 104932090-104932090 on Build GRCh38
rs752531584 Chr.14: 104931990-104931990 on Build GRCh38
rs768388440 Chr.14: 104932127-104932127 on Build GRCh38
rs779386022 Chr.14: 104932020-104932020 on Build GRCh38
rs928173825 Chr.14: 104932156-104932156 on Build GRCh38
rs376735811 Chr.14: 104932102-104932102 on Build GRCh38
rs966594082 Chr.14: 104932115-104932115 on Build GRCh38
rs758884819 Chr.14: 104932066-104932067 on Build GRCh38
rs772840836 Chr.14: 104931973-104931973 on Build GRCh38
rs746653468 Chr.14: 104932107-104932107 on Build GRCh38
rs753210276 Chr.14: 104932161-104932161 on Build GRCh38
rs748586346 Chr.14: 104932021-104932021 on Build GRCh38
rs753267679 Chr.14: 104932082-104932082 on Build GRCh38
rs759019260 Chr.14: 104932144-104932144 on Build GRCh38

View Full Product Details