Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 43644471-43644732 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PLEKHH2
Entrez Gene ID
130271
Gene Name
pleckstrin homology, MyTH4 and FERM domain containing H2
Gene Aliases
PLEKHH1L
UniGene
Hs.164162
Gene Chromosome Location
Chr.2: 43637300-43767987 on Build GRCh38
Transcripts
NM_172069.3 XM_017003352.1 XM_017003353.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1021018 c.47G>A p.R16Q Chr.2: 43644720-43644720 on Build GRCh38
5612447 c.26G>C p.G9A Chr.2: 43644699-43644699 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs374200215 Chr.2: 43644657-43644657 on Build GRCh38
rs779717742 Chr.2: 43644665-43644665 on Build GRCh38
rs757355516 Chr.2: 43644641-43644642 on Build GRCh38
rs188540922 Chr.2: 43644686-43644686 on Build GRCh38
rs35254898 Chr.2: 43644668-43644668 on Build GRCh38
rs749192731 Chr.2: 43644675-43644675 on Build GRCh38
rs558946969 Chr.2: 43644707-43644707 on Build GRCh38
rs910709655 Chr.2: 43644571-43644571 on Build GRCh38
rs1037890536 Chr.2: 43644591-43644591 on Build GRCh38
rs774384857 Chr.2: 43644699-43644699 on Build GRCh38
rs373524206 Chr.2: 43644688-43644688 on Build GRCh38
rs773559760 Chr.2: 43644709-43644709 on Build GRCh38
rs190827037 Chr.2: 43644550-43644550 on Build GRCh38
rs766706430 Chr.2: 43644710-43644710 on Build GRCh38
rs779913532 Chr.2: 43644645-43644648 on Build GRCh38
rs894066988 Chr.2: 43644580-43644580 on Build GRCh38
rs371892737 Chr.2: 43644598-43644598 on Build GRCh38
rs563644047 Chr.2: 43644662-43644662 on Build GRCh38
rs553021967 Chr.2: 43644542-43644542 on Build GRCh38
rs557055910 Chr.2: 43644517-43644517 on Build GRCh38
rs906582410 Chr.2: 43644656-43644656 on Build GRCh38
rs567230029 Chr.2: 43644522-43644522 on Build GRCh38
rs943486678 Chr.2: 43644577-43644577 on Build GRCh38
rs77006940 Chr.2: 43644640-43644640 on Build GRCh38
rs745581411 Chr.2: 43644628-43644628 on Build GRCh38
rs775330078 Chr.2: 43644661-43644661 on Build GRCh38
rs954941363 Chr.2: 43644536-43644536 on Build GRCh38
rs906733011 Chr.2: 43644702-43644702 on Build GRCh38
rs1054407097 Chr.2: 43644565-43644565 on Build GRCh38
rs536290817 Chr.2: 43644539-43644539 on Build GRCh38
rs932374551 Chr.2: 43644551-43644551 on Build GRCh38
rs929414905 Chr.2: 43644634-43644634 on Build GRCh38

View Full Product Details