Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 21777910-21778159 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MIB1
Entrez Gene ID
57534
Gene Name
mindbomb E3 ubiquitin protein ligase 1
Gene Aliases
DIP-1, DIP1, LVNC7, MIB, ZZANK2, ZZZ6
UniGene
Hs.140903
Gene Chromosome Location
Chr.18: 21740793-21870957 on Build GRCh38
Transcripts
NM_020774.3 XM_017025873.1 XM_017025874.1 XM_017025875.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3403457 c.670G>A p.G224S Chr.18: 21778136-21778136 on Build GRCh38
7170201 c.678C>G p.F226L Chr.18: 21778144-21778144 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs745887765 Chr.18: 21778076-21778076 on Build GRCh38
rs757662902 Chr.18: 21778062-21778062 on Build GRCh38
rs140258431 Chr.18: 21778109-21778109 on Build GRCh38
rs987312927 Chr.18: 21777985-21777985 on Build GRCh38
rs77229914 Chr.18: 21778005-21778005 on Build GRCh38
rs752128136 Chr.18: 21778061-21778061 on Build GRCh38
rs771928152 Chr.18: 21778090-21778090 on Build GRCh38
rs369589287 Chr.18: 21778078-21778078 on Build GRCh38
rs760668586 Chr.18: 21778101-21778101 on Build GRCh38
rs145640281 Chr.18: 21778054-21778054 on Build GRCh38
rs770717969 Chr.18: 21778102-21778102 on Build GRCh38
rs765022698 Chr.18: 21778110-21778110 on Build GRCh38
rs113529696 Chr.18: 21778116-21778116 on Build GRCh38
rs986453966 Chr.18: 21778129-21778129 on Build GRCh38
rs747946329 Chr.18: 21778082-21778082 on Build GRCh38
rs775260348 Chr.18: 21778115-21778115 on Build GRCh38
rs1031730633 Chr.18: 21777951-21777951 on Build GRCh38
rs770056880 Chr.18: 21778077-21778077 on Build GRCh38
rs762463590 Chr.18: 21778124-21778124 on Build GRCh38
rs956102948 Chr.18: 21777995-21777995 on Build GRCh38
rs778191320 Chr.18: 21778057-21778057 on Build GRCh38
rs763638129 Chr.18: 21778081-21778082 on Build GRCh38
rs1022578631 Chr.18: 21778031-21778031 on Build GRCh38
rs781250284 Chr.18: 21778074-21778074 on Build GRCh38
rs760209786 Chr.18: 21778069-21778070 on Build GRCh38
rs111601483 Chr.18: 21778091-21778091 on Build GRCh38
rs967901399 Chr.18: 21778053-21778053 on Build GRCh38
rs776340384 Chr.18: 21778106-21778106 on Build GRCh38

View Full Product Details