Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 94378527-94378768 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RAD54B
Entrez Gene ID
25788
Gene Name
RAD54 homolog B (S. cerevisiae)
Gene Aliases
RDH54
UniGene
Hs.30561
Gene Chromosome Location
Chr.8: 94371960-94475115 on Build GRCh38
Transcripts
NM_001205263.1 NM_012415.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4846596 c.2275C>T p.Q759* Chr.8: 94378607-94378607 on Build GRCh38
3651920 c.2284C>T p.P762S Chr.8: 94378598-94378598 on Build GRCh38
4914735 c.2248-2A>G p.? Chr.8: 94378636-94378636 on Build GRCh38
1102561 c.2267G>T p.R756I Chr.8: 94378615-94378615 on Build GRCh38
6357976 c.2300G>A p.R767K Chr.8: 94378582-94378582 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs920269773 Chr.8: 94378591-94378591 on Build GRCh38
rs892828837 Chr.8: 94378738-94378738 on Build GRCh38
rs534302385 Chr.8: 94378690-94378690 on Build GRCh38
rs775851697 Chr.8: 94378695-94378695 on Build GRCh38
rs1045553929 Chr.8: 94378707-94378707 on Build GRCh38
rs746311477 Chr.8: 94378669-94378669 on Build GRCh38
rs941344826 Chr.8: 94378691-94378691 on Build GRCh38
rs716770 Chr.8: 94378693-94378693 on Build GRCh38
rs774940010 Chr.8: 94378666-94378666 on Build GRCh38
rs757913367 Chr.8: 94378609-94378609 on Build GRCh38
rs540736026 Chr.8: 94378731-94378731 on Build GRCh38
rs777515642 Chr.8: 94378709-94378709 on Build GRCh38
rs771402807 Chr.8: 94378657-94378657 on Build GRCh38
rs553190806 Chr.8: 94378683-94378683 on Build GRCh38
rs779468249 Chr.8: 94378612-94378612 on Build GRCh38
rs929858498 Chr.8: 94378700-94378700 on Build GRCh38
rs759679204 Chr.8: 94378563-94378563 on Build GRCh38
rs765216364 Chr.8: 94378581-94378581 on Build GRCh38
rs749662439 Chr.8: 94378651-94378651 on Build GRCh38
rs774365453 Chr.8: 94378560-94378560 on Build GRCh38
rs753645144 Chr.8: 94378575-94378575 on Build GRCh38
rs374100160 Chr.8: 94378572-94378572 on Build GRCh38
rs147396183 Chr.8: 94378629-94378629 on Build GRCh38
rs771440070 Chr.8: 94378559-94378559 on Build GRCh38
rs749272275 Chr.8: 94378725-94378725 on Build GRCh38
rs772309341 Chr.8: 94378672-94378672 on Build GRCh38
rs746692746 Chr.8: 94378626-94378626 on Build GRCh38
rs371550113 Chr.8: 94378684-94378684 on Build GRCh38
rs539641872 Chr.8: 94378682-94378682 on Build GRCh38
rs767648288 Chr.8: 94378565-94378565 on Build GRCh38
rs935927888 Chr.8: 94378736-94378736 on Build GRCh38
rs368086541 Chr.8: 94378638-94378638 on Build GRCh38
rs775859629 Chr.8: 94378570-94378570 on Build GRCh38
rs369247947 Chr.8: 94378733-94378733 on Build GRCh38
rs757180812 Chr.8: 94378580-94378580 on Build GRCh38
rs749959340 Chr.8: 94378592-94378592 on Build GRCh38
rs893852884 Chr.8: 94378740-94378740 on Build GRCh38

View Full Product Details