Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 32927960-32928193 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
QSER1
Entrez Gene ID
79832
Gene Name
glutamine and serine rich 1
Gene Aliases
-
UniGene
Hs.369368
Gene Chromosome Location
Chr.11: 32892826-32980270 on Build GRCh38
Transcripts
NM_001076786.2 XM_006718325.3 XM_017018327.1 XM_017018328.1 XM_017018329.1 XM_017018330.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7111338 c.93G>A p.Q31Q Chr.11: 32928119-32928119 on Build GRCh38
6318562 c.34G>T p.A12S Chr.11: 32928060-32928060 on Build GRCh38
6505140 c.61C>T p.L21F Chr.11: 32928087-32928087 on Build GRCh38
926407 c.23A>G p.E8G Chr.11: 32928049-32928049 on Build GRCh38
3446818 c.63C>T p.L21L Chr.11: 32928089-32928089 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs751942678 Chr.11: 32928079-32928079 on Build GRCh38
rs902559571 Chr.11: 32928085-32928085 on Build GRCh38
rs763490135 Chr.11: 32928058-32928058 on Build GRCh38
rs201919670 Chr.11: 32928066-32928066 on Build GRCh38
rs542781649 Chr.11: 32928140-32928140 on Build GRCh38
rs756209033 Chr.11: 32928100-32928100 on Build GRCh38
rs1032726690 Chr.11: 32928137-32928137 on Build GRCh38
rs764779354 Chr.11: 32928042-32928042 on Build GRCh38
rs529272123 Chr.11: 32928120-32928120 on Build GRCh38
rs760293291 Chr.11: 32928029-32928029 on Build GRCh38
rs746814550 Chr.11: 32928129-32928129 on Build GRCh38
rs761505426 Chr.11: 32928156-32928156 on Build GRCh38
rs781398418 Chr.11: 32928082-32928082 on Build GRCh38
rs768408915 Chr.11: 32928032-32928032 on Build GRCh38
rs745833150 Chr.11: 32928119-32928119 on Build GRCh38
rs1028060839 Chr.11: 32928108-32928108 on Build GRCh38
rs772760038 Chr.11: 32928165-32928165 on Build GRCh38
rs755224358 Chr.11: 32928080-32928080 on Build GRCh38
rs775323163 Chr.11: 32928012-32928012 on Build GRCh38
rs745770195 Chr.11: 32927997-32927997 on Build GRCh38
rs201340164 Chr.11: 32928121-32928121 on Build GRCh38
rs540486719 Chr.11: 32928057-32928057 on Build GRCh38
rs578007156 Chr.11: 32928055-32928055 on Build GRCh38
rs752860283 Chr.11: 32928084-32928084 on Build GRCh38
rs544419749 Chr.11: 32927999-32927999 on Build GRCh38
rs768496938 Chr.11: 32928138-32928138 on Build GRCh38
rs201338306 Chr.11: 32928161-32928161 on Build GRCh38
rs200211395 Chr.11: 32928038-32928038 on Build GRCh38
rs761409897 Chr.11: 32928040-32928040 on Build GRCh38
rs777791072 Chr.11: 32928111-32928111 on Build GRCh38

View Full Product Details