Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 6026139-6026378 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
JAKMIP1
Entrez Gene ID
152789
Gene Name
janus kinase and microtubule interacting protein 1
Gene Aliases
Gababrbp, JAMIP1, MARLIN1
UniGene
Hs.479066
Gene Chromosome Location
Chr.4: 6024925-6200591 on Build GRCh38
Transcripts
NM_001099433.1 XM_017007788.1 XM_017007789.1 XM_017007790.1 XM_017007791.1 XM_017007792.1 XM_017007793.1 XM_017007794.1 XM_017007795.1 XM_017007797.1 XM_017007798.1 XM_017007799.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3302527 c.2462delT p.L821fs*5 Chr.4: 6026262-6026262 on Build GRCh38
5401372 c.2455C>T p.L819F Chr.4: 6026269-6026269 on Build GRCh38
5026121 c.2485C>T p.L829L Chr.4: 6026239-6026239 on Build GRCh38
1430392 c.2462_2463insT p.L821fs*>12 Chr.4: 6026261-6026262 on Build GRCh38
6236420 c.2471C>A p.S824* Chr.4: 6026253-6026253 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs755687589 Chr.4: 6026204-6026204 on Build GRCh38
rs778212826 Chr.4: 6026307-6026308 on Build GRCh38
rs201451956 Chr.4: 6026180-6026180 on Build GRCh38
rs902961856 Chr.4: 6026283-6026283 on Build GRCh38
rs902983335 Chr.4: 6026337-6026337 on Build GRCh38
rs550590500 Chr.4: 6026219-6026219 on Build GRCh38
rs79962349 Chr.4: 6026261-6026261 on Build GRCh38
rs749038820 Chr.4: 6026227-6026227 on Build GRCh38
rs564914875 Chr.4: 6026182-6026182 on Build GRCh38
rs772410033 Chr.4: 6026205-6026205 on Build GRCh38
rs953101483 Chr.4: 6026161-6026162 on Build GRCh38
rs535467405 Chr.4: 6026167-6026167 on Build GRCh38
rs770097759 Chr.4: 6026183-6026183 on Build GRCh38
rs372177265 Chr.4: 6026207-6026207 on Build GRCh38
rs1050213002 Chr.4: 6026215-6026215 on Build GRCh38
rs1037562834 Chr.4: 6026179-6026179 on Build GRCh38
rs747979500 Chr.4: 6026311-6026311 on Build GRCh38
rs778270565 Chr.4: 6026185-6026185 on Build GRCh38
rs765657292 Chr.4: 6026171-6026171 on Build GRCh38
rs183884928 Chr.4: 6026199-6026199 on Build GRCh38
rs893824611 Chr.4: 6026249-6026249 on Build GRCh38
rs947261823 Chr.4: 6026272-6026272 on Build GRCh38
rs559890400 Chr.4: 6026306-6026306 on Build GRCh38

View Full Product Details