Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 170870036-170870291 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RPL22L1
Entrez Gene ID
200916
Gene Name
ribosomal protein L22 like 1
Gene Aliases
-
UniGene
Hs.380933
Gene Chromosome Location
Chr.3: 170864875-170870256 on Build GRCh38
Transcripts
NM_001099645.1 NM_001320451.1 NR_135259.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7116077 c.8C>T p.P3L Chr.3: 170870160-170870160 on Build GRCh38
4564008 c.9G>T p.P3P Chr.3: 170870159-170870159 on Build GRCh38
6163952 c.3G>T p.M1I Chr.3: 170870165-170870165 on Build GRCh38
1327728 c.7C>T p.P3S Chr.3: 170870161-170870161 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs554710245 Chr.3: 170870166-170870166 on Build GRCh38
rs762805405 Chr.3: 170870253-170870253 on Build GRCh38
rs748719784 Chr.3: 170870216-170870216 on Build GRCh38
rs777034073 Chr.3: 170870181-170870181 on Build GRCh38
rs948568062 Chr.3: 170870243-170870243 on Build GRCh38
rs763418664 Chr.3: 170870190-170870190 on Build GRCh38
rs751661574 Chr.3: 170870161-170870161 on Build GRCh38
rs750615835 Chr.3: 170870153-170870153 on Build GRCh38
rs1036427211 Chr.3: 170870237-170870237 on Build GRCh38
rs372204393 Chr.3: 170870144-170870144 on Build GRCh38
rs559938018 Chr.3: 170870267-170870267 on Build GRCh38
rs778606198 Chr.3: 170870167-170870167 on Build GRCh38
rs762255208 Chr.3: 170870179-170870179 on Build GRCh38
rs755099149 Chr.3: 170870157-170870157 on Build GRCh38
rs199670825 Chr.3: 170870128-170870128 on Build GRCh38
rs996002056 Chr.3: 170870253-170870273 on Build GRCh38
rs200784037 Chr.3: 170870160-170870160 on Build GRCh38
rs530360967 Chr.3: 170870209-170870209 on Build GRCh38
rs994048576 Chr.3: 170870257-170870257 on Build GRCh38
rs908183020 Chr.3: 170870207-170870207 on Build GRCh38
rs766708802 Chr.3: 170870191-170870191 on Build GRCh38
rs765519693 Chr.3: 170870152-170870152 on Build GRCh38
rs775654994 Chr.3: 170870206-170870206 on Build GRCh38
rs745723245 Chr.3: 170870168-170870168 on Build GRCh38
rs369828515 Chr.3: 170870266-170870266 on Build GRCh38
rs942135611 Chr.3: 170870226-170870226 on Build GRCh38
rs376598533 Chr.3: 170870211-170870211 on Build GRCh38
rs938467215 Chr.3: 170870222-170870222 on Build GRCh38
rs563175620 Chr.3: 170870204-170870204 on Build GRCh38
rs371396509 Chr.3: 170870171-170870171 on Build GRCh38
rs540334886 Chr.3: 170870194-170870194 on Build GRCh38
rs376190297 Chr.3: 170870260-170870260 on Build GRCh38
rs544475773 Chr.3: 170870203-170870203 on Build GRCh38
rs778947962 Chr.3: 170870197-170870197 on Build GRCh38
rs760216349 Chr.3: 170870130-170870130 on Build GRCh38
rs994470496 Chr.3: 170870100-170870100 on Build GRCh38
rs762176410 Chr.3: 170870149-170870149 on Build GRCh38
rs748123968 Chr.3: 170870190-170870191 on Build GRCh38
rs758270443 Chr.3: 170870202-170870202 on Build GRCh38
rs755179925 Chr.3: 170870164-170870164 on Build GRCh38
rs757166528 Chr.3: 170870196-170870196 on Build GRCh38
rs770172126 Chr.3: 170870210-170870210 on Build GRCh38
rs915768793 Chr.3: 170870241-170870241 on Build GRCh38
rs771592478 Chr.3: 170870170-170870170 on Build GRCh38
rs985181044 Chr.3: 170870159-170870159 on Build GRCh38
rs577040119 Chr.3: 170870199-170870199 on Build GRCh38
rs777051933 Chr.3: 170870173-170870173 on Build GRCh38
rs777180241 Chr.3: 170870147-170870147 on Build GRCh38
rs573287184 Chr.3: 170870193-170870193 on Build GRCh38
rs367602421 Chr.3: 170870163-170870163 on Build GRCh38
rs369862481 Chr.3: 170870195-170870195 on Build GRCh38
rs768165142 Chr.3: 170870172-170870172 on Build GRCh38
rs372926652 Chr.3: 170870208-170870208 on Build GRCh38
rs745514104 Chr.3: 170870083-170870083 on Build GRCh38
rs771572724 Chr.3: 170870120-170870120 on Build GRCh38
rs375274315 Chr.3: 170870188-170870188 on Build GRCh38
rs770016761 Chr.3: 170870198-170870198 on Build GRCh38
rs753841119 Chr.3: 170870165-170870165 on Build GRCh38

View Full Product Details