Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 107057234-107057487 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PRMT6
Entrez Gene ID
55170
Gene Name
protein arginine methyltransferase 6
Gene Aliases
HRMT1L6
UniGene
Hs.26006
Gene Chromosome Location
Chr.1: 107056645-107059294 on Build GRCh38
Transcripts
NM_018137.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2116376 c.670G>T p.G224C Chr.1: 107057385-107057385 on Build GRCh38
6756301 c.674T>A p.F225Y Chr.1: 107057389-107057389 on Build GRCh38
4614643 c.352_353insT p.R119fs*32 Chr.1: 107057387-107057388 on Build GRCh38
2116377 c.493G>T p.G165C Chr.1: 107057385-107057385 on Build GRCh38
6364527 c.405C>T p.V135V Chr.1: 107057440-107057440 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs759568299 Chr.1: 107057410-107057410 on Build GRCh38
rs749308282 Chr.1: 107057452-107057452 on Build GRCh38
rs2232016 Chr.1: 107057296-107057296 on Build GRCh38
rs554348171 Chr.1: 107057444-107057444 on Build GRCh38
rs765270343 Chr.1: 107057429-107057429 on Build GRCh38
rs765679967 Chr.1: 107057292-107057292 on Build GRCh38
rs778175587 Chr.1: 107057300-107057300 on Build GRCh38
rs773355841 Chr.1: 107057270-107057270 on Build GRCh38
rs376689218 Chr.1: 107057282-107057282 on Build GRCh38
rs745323593 Chr.1: 107057346-107057346 on Build GRCh38
rs943676790 Chr.1: 107057299-107057299 on Build GRCh38
rs777418693 Chr.1: 107057361-107057361 on Build GRCh38
rs776878092 Chr.1: 107057464-107057464 on Build GRCh38
rs372142027 Chr.1: 107057450-107057450 on Build GRCh38
rs369781877 Chr.1: 107057267-107057267 on Build GRCh38
rs374379636 Chr.1: 107057289-107057289 on Build GRCh38
rs75744254 Chr.1: 107057336-107057336 on Build GRCh38
rs946243301 Chr.1: 107057420-107057420 on Build GRCh38
rs766561555 Chr.1: 107057395-107057395 on Build GRCh38
rs145591038 Chr.1: 107057441-107057441 on Build GRCh38
rs755864626 Chr.1: 107057348-107057348 on Build GRCh38
rs776710014 Chr.1: 107057401-107057401 on Build GRCh38
rs1019987606 Chr.1: 107057418-107057418 on Build GRCh38
rs749220337 Chr.1: 107057362-107057362 on Build GRCh38
rs747924265 Chr.1: 107057460-107057460 on Build GRCh38
rs1045684434 Chr.1: 107057295-107057295 on Build GRCh38
rs374493151 Chr.1: 107057272-107057272 on Build GRCh38
rs774027876 Chr.1: 107057454-107057454 on Build GRCh38
rs779702003 Chr.1: 107057451-107057451 on Build GRCh38
rs753806049 Chr.1: 107057347-107057348 on Build GRCh38
rs374565859 Chr.1: 107057453-107057453 on Build GRCh38
rs377462998 Chr.1: 107057258-107057258 on Build GRCh38
rs1055860282 Chr.1: 107057468-107057468 on Build GRCh38
rs371259459 Chr.1: 107057288-107057288 on Build GRCh38
rs759554315 Chr.1: 107057257-107057257 on Build GRCh38
rs753250896 Chr.1: 107057431-107057431 on Build GRCh38
rs367705333 Chr.1: 107057443-107057443 on Build GRCh38
rs746697031 Chr.1: 107057311-107057311 on Build GRCh38
rs756869283 Chr.1: 107057319-107057319 on Build GRCh38
rs745485619 Chr.1: 107057445-107057445 on Build GRCh38
rs777901002 Chr.1: 107057265-107057267 on Build GRCh38
rs1009963665 Chr.1: 107057373-107057373 on Build GRCh38
rs557473664 Chr.1: 107057264-107057264 on Build GRCh38
rs892957203 Chr.1: 107057366-107057366 on Build GRCh38
rs774194962 Chr.1: 107057383-107057383 on Build GRCh38
rs575093469 Chr.1: 107057439-107057439 on Build GRCh38
rs775612422 Chr.1: 107057354-107057354 on Build GRCh38
rs763918627 Chr.1: 107057268-107057268 on Build GRCh38
rs768396085 Chr.1: 107057365-107057365 on Build GRCh38
rs769883942 Chr.1: 107057353-107057353 on Build GRCh38
rs370615431 Chr.1: 107057387-107057387 on Build GRCh38
rs771085775 Chr.1: 107057463-107057463 on Build GRCh38

View Full Product Details