Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 232806156-232806427 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MAP10
Entrez Gene ID
54627
Gene Name
microtubule associated protein 10
Gene Aliases
KIAA1383, MTR120
UniGene
Hs.160373
Gene Chromosome Location
Chr.1: 232804892-232810346 on Build GRCh38
Transcripts
NM_019090.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3677606 c.1314C>A p.T438T Chr.1: 232806337-232806337 on Build GRCh38
5694417 c.1320_1340del21 p.F441_Y447delFCPPPLY Chr.1: 232806343-232806363 on Build GRCh38
3667481 c.1332T>A p.P444P Chr.1: 232806355-232806355 on Build GRCh38
3677608 c.1356C>A p.T452T Chr.1: 232806379-232806379 on Build GRCh38
7110297 c.1278C>A p.S426S Chr.1: 232806301-232806301 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs371993579 Chr.1: 232806393-232806393 on Build GRCh38
rs758393117 Chr.1: 232806258-232806258 on Build GRCh38
rs769974269 Chr.1: 232806228-232806228 on Build GRCh38
rs770972855 Chr.1: 232806363-232806363 on Build GRCh38
rs1054650358 Chr.1: 232806257-232806257 on Build GRCh38
rs746089137 Chr.1: 232806372-232806372 on Build GRCh38
rs776134385 Chr.1: 232806255-232806255 on Build GRCh38
rs777230519 Chr.1: 232806392-232806392 on Build GRCh38
rs368167890 Chr.1: 232806391-232806391 on Build GRCh38
rs775734557 Chr.1: 232806388-232806388 on Build GRCh38
rs754884715 Chr.1: 232806321-232806321 on Build GRCh38
rs761076035 Chr.1: 232806266-232806266 on Build GRCh38
rs560481621 Chr.1: 232806222-232806222 on Build GRCh38
rs561354519 Chr.1: 232806198-232806205 on Build GRCh38
rs767678428 Chr.1: 232806202-232806205 on Build GRCh38
rs780194188 Chr.1: 232806284-232806284 on Build GRCh38
rs531801115 Chr.1: 232806367-232806367 on Build GRCh38
rs765980497 Chr.1: 232806394-232806394 on Build GRCh38
rs766153122 Chr.1: 232806300-232806300 on Build GRCh38
rs373778338 Chr.1: 232806229-232806229 on Build GRCh38
rs777722525 Chr.1: 232806356-232806356 on Build GRCh38
rs772295767 Chr.1: 232806252-232806252 on Build GRCh38
rs368509387 Chr.1: 232806285-232806285 on Build GRCh38
rs372138834 Chr.1: 232806351-232806351 on Build GRCh38
rs775704540 Chr.1: 232806274-232806275 on Build GRCh38
rs761536718 Chr.1: 232806374-232806375 on Build GRCh38
rs764608230 Chr.1: 232806279-232806279 on Build GRCh38
rs749295617 Chr.1: 232806243-232806243 on Build GRCh38
rs371325479 Chr.1: 232806247-232806247 on Build GRCh38
rs1041659527 Chr.1: 232806348-232806348 on Build GRCh38
rs774075737 Chr.1: 232806337-232806337 on Build GRCh38
rs1051817655 Chr.1: 232806185-232806185 on Build GRCh38
rs772549229 Chr.1: 232806375-232806375 on Build GRCh38
rs200508444 Chr.1: 232806353-232806353 on Build GRCh38
rs373605405 Chr.1: 232806182-232806182 on Build GRCh38
rs376618811 Chr.1: 232806204-232806204 on Build GRCh38
rs762513237 Chr.1: 232806290-232806290 on Build GRCh38
rs190242642 Chr.1: 232806187-232806187 on Build GRCh38
rs181890143 Chr.1: 232806328-232806328 on Build GRCh38
rs370663103 Chr.1: 232806211-232806211 on Build GRCh38
rs759940347 Chr.1: 232806250-232806250 on Build GRCh38
rs774504473 Chr.1: 232806249-232806249 on Build GRCh38
rs367589222 Chr.1: 232806245-232806245 on Build GRCh38
rs772323797 Chr.1: 232806358-232806358 on Build GRCh38
rs61739198 Chr.1: 232806282-232806282 on Build GRCh38
rs549361795 Chr.1: 232806316-232806316 on Build GRCh38
rs749389162 Chr.1: 232806357-232806357 on Build GRCh38
rs918903527 Chr.1: 232806343-232806344 on Build GRCh38

View Full Product Details