Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 59365201-59365432 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
OR5AN1
Entrez Gene ID
390195
Gene Name
olfactory receptor family 5 subfamily AN member 1
Gene Aliases
OR11-244
UniGene
Hs.553755
Gene Chromosome Location
Chr.11: 59364459-59365394 on Build GRCh38
Transcripts
NM_001004729.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6201317 c.915G>T p.L305F Chr.11: 59365373-59365373 on Build GRCh38
7103272 c.753C>G p.L251L Chr.11: 59365211-59365211 on Build GRCh38
5480203 c.901G>T p.A301S Chr.11: 59365359-59365359 on Build GRCh38
5081263 c.844G>T p.V282F Chr.11: 59365302-59365302 on Build GRCh38
3687404 c.914T>C p.L305S Chr.11: 59365372-59365372 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs201961211 Chr.11: 59365259-59365259 on Build GRCh38
rs945205572 Chr.11: 59365396-59365396 on Build GRCh38
rs757140731 Chr.11: 59365358-59365358 on Build GRCh38
rs778642391 Chr.11: 59365281-59365281 on Build GRCh38
rs766898098 Chr.11: 59365235-59365235 on Build GRCh38
rs143949956 Chr.11: 59365390-59365390 on Build GRCh38
rs376445040 Chr.11: 59365340-59365340 on Build GRCh38
rs35155155 Chr.11: 59365396-59365396 on Build GRCh38
rs752926773 Chr.11: 59365339-59365339 on Build GRCh38
rs774808179 Chr.11: 59365324-59365324 on Build GRCh38
rs756754946 Chr.11: 59365258-59365258 on Build GRCh38
rs140118808 Chr.11: 59365385-59365385 on Build GRCh38
rs768109824 Chr.11: 59365293-59365293 on Build GRCh38
rs771506013 Chr.11: 59365389-59365389 on Build GRCh38
rs573328426 Chr.11: 59365247-59365247 on Build GRCh38
rs773835338 Chr.11: 59365295-59365295 on Build GRCh38
rs10896964 Chr.11: 59365409-59365409 on Build GRCh38
rs753954265 Chr.11: 59365345-59365345 on Build GRCh38
rs7941190 Chr.11: 59365325-59365325 on Build GRCh38
rs763593259 Chr.11: 59365297-59365297 on Build GRCh38
rs764202357 Chr.11: 59365343-59365343 on Build GRCh38
rs985278197 Chr.11: 59365296-59365296 on Build GRCh38
rs746524691 Chr.11: 59365286-59365286 on Build GRCh38
rs752092477 Chr.11: 59365284-59365285 on Build GRCh38
rs768178926 Chr.11: 59365381-59365381 on Build GRCh38
rs146617781 Chr.11: 59365366-59365366 on Build GRCh38
rs746691252 Chr.11: 59365379-59365379 on Build GRCh38
rs182721482 Chr.11: 59365261-59365261 on Build GRCh38
rs767708209 Chr.11: 59365332-59365332 on Build GRCh38
rs759936814 Chr.11: 59365402-59365402 on Build GRCh38
rs767023034 Chr.11: 59365303-59365303 on Build GRCh38
rs1034375597 Chr.11: 59365373-59365373 on Build GRCh38
rs551614545 Chr.11: 59365363-59365363 on Build GRCh38
rs370394618 Chr.11: 59365374-59365374 on Build GRCh38
rs757017833 Chr.11: 59365269-59365269 on Build GRCh38
rs752761602 Chr.11: 59365255-59365255 on Build GRCh38
rs778838873 Chr.11: 59365359-59365359 on Build GRCh38
rs751798609 Chr.11: 59365239-59365239 on Build GRCh38
rs755186708 Chr.11: 59365242-59365242 on Build GRCh38
rs758194458 Chr.11: 59365370-59365370 on Build GRCh38
rs373581022 Chr.11: 59365282-59365282 on Build GRCh38
rs775010238 Chr.11: 59365283-59365283 on Build GRCh38

View Full Product Details