Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 156160340-156160613 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SEMA4A
Entrez Gene ID
64218
Gene Name
semaphorin 4A
Gene Aliases
CORD10, RP35, SEMAB, SEMB
UniGene
Hs.408846
Gene Chromosome Location
Chr.1: 156149673-156177751 on Build GRCh38
Transcripts
NM_001193300.1 NM_001193301.1 NM_001193302.1 NM_022367.3 XM_011509871.2 XM_011509872.2 XM_011509873.2 XM_011509874.2 XM_011509875.2 XM_011509876.2 XM_011509878.2 XM_011509879.2 XM_017002056.1 XM_017002057.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5936942 c.569-6C>T p.? Chr.1: 156160437-156160437 on Build GRCh38
7189178 c.649G>A p.V217I Chr.1: 156160523-156160523 on Build GRCh38
897124 c.588T>C p.G196G Chr.1: 156160462-156160462 on Build GRCh38
4023194 c.673C>T p.R225C Chr.1: 156160547-156160547 on Build GRCh38
5794409 c.586G>C p.G196R Chr.1: 156160460-156160460 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs775604376 Chr.1: 156160528-156160528 on Build GRCh38
rs764259200 Chr.1: 156160539-156160539 on Build GRCh38
rs768335882 Chr.1: 156160531-156160531 on Build GRCh38
rs1008385344 Chr.1: 156160489-156160489 on Build GRCh38
rs779104824 Chr.1: 156160426-156160426 on Build GRCh38
rs1013440701 Chr.1: 156160587-156160587 on Build GRCh38
rs775379713 Chr.1: 156160567-156160567 on Build GRCh38
rs772824102 Chr.1: 156160481-156160481 on Build GRCh38
rs760954058 Chr.1: 156160413-156160413 on Build GRCh38
rs754201054 Chr.1: 156160417-156160417 on Build GRCh38
rs887202270 Chr.1: 156160477-156160477 on Build GRCh38
rs761889239 Chr.1: 156160576-156160576 on Build GRCh38
rs1033393125 Chr.1: 156160577-156160577 on Build GRCh38
rs563967794 Chr.1: 156160361-156160361 on Build GRCh38
rs767596583 Chr.1: 156160406-156160406 on Build GRCh38
rs531400191 Chr.1: 156160438-156160438 on Build GRCh38
rs750958745 Chr.1: 156160430-156160430 on Build GRCh38
rs760718249 Chr.1: 156160537-156160537 on Build GRCh38
rs750752916 Chr.1: 156160591-156160591 on Build GRCh38
rs769669648 Chr.1: 156160395-156160395 on Build GRCh38
rs747933883 Chr.1: 156160468-156160468 on Build GRCh38
rs760661651 Chr.1: 156160585-156160585 on Build GRCh38
rs770850290 Chr.1: 156160516-156160516 on Build GRCh38
rs1041986577 Chr.1: 156160463-156160463 on Build GRCh38
rs745337542 Chr.1: 156160424-156160426 on Build GRCh38
rs758921192 Chr.1: 156160435-156160435 on Build GRCh38
rs890649984 Chr.1: 156160410-156160410 on Build GRCh38
rs764286415 Chr.1: 156160414-156160414 on Build GRCh38
rs752705092 Chr.1: 156160408-156160408 on Build GRCh38
rs771519010 Chr.1: 156160500-156160501 on Build GRCh38
rs762838301 Chr.1: 156160401-156160401 on Build GRCh38
rs769664463 Chr.1: 156160476-156160476 on Build GRCh38
rs780876890 Chr.1: 156160467-156160467 on Build GRCh38
rs773171889 Chr.1: 156160396-156160396 on Build GRCh38
rs765650353 Chr.1: 156160590-156160590 on Build GRCh38
rs202199807 Chr.1: 156160429-156160429 on Build GRCh38
rs958834880 Chr.1: 156160580-156160580 on Build GRCh38
rs1009009521 Chr.1: 156160453-156160453 on Build GRCh38
rs145701501 Chr.1: 156160450-156160450 on Build GRCh38
rs371388874 Chr.1: 156160498-156160498 on Build GRCh38
rs754739313 Chr.1: 156160466-156160466 on Build GRCh38
rs776940811 Chr.1: 156160559-156160559 on Build GRCh38
rs1024379954 Chr.1: 156160588-156160588 on Build GRCh38

View Full Product Details