Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 216859490-216859749 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TNP1
Entrez Gene ID
7141
Gene Name
transition protein 1
Gene Aliases
TP1
UniGene
Hs.3017
Gene Chromosome Location
Chr.2: 216859458-216860059 on Build GRCh38
Transcripts
NM_003284.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3577585 c.158C>T p.S53F Chr.2: 216859677-216859677 on Build GRCh38
291186 c.154C>T p.R52C Chr.2: 216859681-216859681 on Build GRCh38
5097400 c.147C>T p.R49R Chr.2: 216859688-216859688 on Build GRCh38
442297 c.158C>G p.S53C Chr.2: 216859677-216859677 on Build GRCh38
6805470 c.155G>A p.R52H Chr.2: 216859680-216859680 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs761892156 Chr.2: 216859673-216859673 on Build GRCh38
rs760854000 Chr.2: 216859687-216859687 on Build GRCh38
rs1038703973 Chr.2: 216859617-216859617 on Build GRCh38
rs201394603 Chr.2: 216859681-216859681 on Build GRCh38
rs368014196 Chr.2: 216859653-216859653 on Build GRCh38
rs756861746 Chr.2: 216859711-216859711 on Build GRCh38
rs911683540 Chr.2: 216859584-216859584 on Build GRCh38
rs370918270 Chr.2: 216859658-216859658 on Build GRCh38
rs374598465 Chr.2: 216859640-216859640 on Build GRCh38
rs549809988 Chr.2: 216859654-216859654 on Build GRCh38
rs549146459 Chr.2: 216859523-216859523 on Build GRCh38
rs758063596 Chr.2: 216859693-216859693 on Build GRCh38
rs776449743 Chr.2: 216859657-216859657 on Build GRCh38
rs142667039 Chr.2: 216859692-216859692 on Build GRCh38
rs200357825 Chr.2: 216859703-216859703 on Build GRCh38
rs957005652 Chr.2: 216859592-216859592 on Build GRCh38
rs562802006 Chr.2: 216859661-216859661 on Build GRCh38
rs748074741 Chr.2: 216859638-216859638 on Build GRCh38
rs746534573 Chr.2: 216859648-216859648 on Build GRCh38
rs890578284 Chr.2: 216859544-216859544 on Build GRCh38
rs879649353 Chr.2: 216859680-216859680 on Build GRCh38
rs374305305 Chr.2: 216859582-216859582 on Build GRCh38
rs754585675 Chr.2: 216859721-216859722 on Build GRCh38
rs955552533 Chr.2: 216859581-216859581 on Build GRCh38
rs775905315 Chr.2: 216859660-216859660 on Build GRCh38
rs778638482 Chr.2: 216859627-216859627 on Build GRCh38
rs761974902 Chr.2: 216859708-216859708 on Build GRCh38
rs763425353 Chr.2: 216859664-216859664 on Build GRCh38
rs538074565 Chr.2: 216859594-216859594 on Build GRCh38
rs969035528 Chr.2: 216859644-216859644 on Build GRCh38
rs553835924 Chr.2: 216859689-216859689 on Build GRCh38
rs754789346 Chr.2: 216859622-216859622 on Build GRCh38
rs147713572 Chr.2: 216859670-216859670 on Build GRCh38
rs1048134051 Chr.2: 216859686-216859686 on Build GRCh38
rs780822708 Chr.2: 216859715-216859715 on Build GRCh38
rs745566654 Chr.2: 216859719-216859719 on Build GRCh38
rs996294515 Chr.2: 216859522-216859522 on Build GRCh38
rs943061154 Chr.2: 216859600-216859600 on Build GRCh38
rs369895229 Chr.2: 216859641-216859641 on Build GRCh38
rs777779863 Chr.2: 216859695-216859695 on Build GRCh38
rs908437904 Chr.2: 216859694-216859694 on Build GRCh38
rs924196535 Chr.2: 216859546-216859546 on Build GRCh38

View Full Product Details