Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 32473922-32474147 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC7
Entrez Gene ID
79741
Gene Name
coiled-coil domain containing 7
Gene Aliases
BIOT2, BioT2-A, BioT2-B, BioT2-C, C10orf68
UniGene
Hs.585464
Gene Chromosome Location
Chr.10: 32446082-32882864 on Build GRCh38
Transcripts
NM_001026383.2 NM_001321115.1 NM_145023.5 NR_109826.1 NR_109827.1 XM_006717505.1 XM_006717506.1 XM_011519663.1 XM_011519664.1 XM_011519666.1 XM_011519667.1 XM_011519668.1 XM_011519669.1 XM_011519670.1 XM_011519671.1 XM_011519672.1 XM_011519673.1 XM_011519674.1 XM_011519675.1 XM_011519676.1 XM_011519677.1 XM_011519678.1 XM_011519679.1 XM_011519680.2 XM_011519684.2 XM_011519685.1 XM_011519686.2 XM_011519688.2 XM_017016638.1 XM_017016639.1 XM_017016640.1 XM_017016641.1 XM_017016642.1 XM_017016643.1 XM_017016644.1 XM_017016645.1 XM_017016647.1 XM_017016650.1 XM_017016651.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4626644 c.780T>C p.V260V Chr.10: 32474007-32474007 on Build GRCh38
159808 c.758C>G p.S253* Chr.10: 32473985-32473985 on Build GRCh38
1627475 c.796C>T p.Q266* Chr.10: 32474023-32474023 on Build GRCh38
5677039 c.778G>A p.V260I Chr.10: 32474005-32474005 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs758035816 Chr.10: 32474028-32474028 on Build GRCh38
rs765118327 Chr.10: 32474094-32474094 on Build GRCh38
rs145224620 Chr.10: 32473981-32473981 on Build GRCh38
rs771439165 Chr.10: 32474025-32474025 on Build GRCh38
rs748512643 Chr.10: 32474015-32474015 on Build GRCh38
rs552663297 Chr.10: 32474120-32474120 on Build GRCh38
rs370404173 Chr.10: 32474048-32474048 on Build GRCh38
rs886301419 Chr.10: 32474119-32474119 on Build GRCh38
rs774998825 Chr.10: 32474029-32474029 on Build GRCh38
rs747468492 Chr.10: 32473988-32473988 on Build GRCh38
rs761488606 Chr.10: 32474092-32474092 on Build GRCh38
rs150437631 Chr.10: 32474024-32474024 on Build GRCh38
rs753827377 Chr.10: 32474071-32474071 on Build GRCh38
rs760535162 Chr.10: 32474035-32474035 on Build GRCh38
rs112534766 Chr.10: 32474018-32474018 on Build GRCh38
rs200004319 Chr.10: 32474043-32474043 on Build GRCh38
rs750759638 Chr.10: 32474083-32474083 on Build GRCh38
rs750705104 Chr.10: 32473978-32473978 on Build GRCh38
rs770244228 Chr.10: 32474016-32474016 on Build GRCh38
rs780311880 Chr.10: 32473985-32473985 on Build GRCh38
rs778986372 Chr.10: 32473977-32473977 on Build GRCh38
rs757139670 Chr.10: 32473973-32473973 on Build GRCh38
rs1019770668 Chr.10: 32473998-32473998 on Build GRCh38
rs150995497 Chr.10: 32474003-32474003 on Build GRCh38
rs201948918 Chr.10: 32474023-32474023 on Build GRCh38
rs140485770 Chr.10: 32474014-32474014 on Build GRCh38
rs777472299 Chr.10: 32474063-32474064 on Build GRCh38

View Full Product Details