Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 54811857-54812088 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LEXM
Entrez Gene ID
163747
Gene Name
lymphocyte expansion molecule
Gene Aliases
C1orf177, LEM
UniGene
Hs.376018
Gene Chromosome Location
Chr.1: 54806063-54842264 on Build GRCh38
Transcripts
NM_001110533.1 NM_152607.2 XM_017000480.1 XM_017000481.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3491009 c.568G>A p.E190K Chr.1: 54811881-54811881 on Build GRCh38
5554919 c.637G>A p.G213S Chr.1: 54812064-54812064 on Build GRCh38
4554046 c.609G>A p.R203R Chr.1: 54811922-54811922 on Build GRCh38
3491010 c.571G>A p.G191S Chr.1: 54811884-54811884 on Build GRCh38
7358278 c.617A>G p.Q206R Chr.1: 54811930-54811930 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs200756384 Chr.1: 54811938-54811938 on Build GRCh38
rs560071108 Chr.1: 54811921-54811921 on Build GRCh38
rs1011055937 Chr.1: 54812035-54812035 on Build GRCh38
rs763928879 Chr.1: 54811983-54811983 on Build GRCh38
rs753453051 Chr.1: 54811907-54811907 on Build GRCh38
rs755993469 Chr.1: 54811980-54811981 on Build GRCh38
rs114564449 Chr.1: 54812008-54812008 on Build GRCh38
rs199976702 Chr.1: 54811968-54811968 on Build GRCh38
rs927648503 Chr.1: 54811900-54811900 on Build GRCh38
rs777946545 Chr.1: 54812037-54812049 on Build GRCh38
rs770770079 Chr.1: 54811953-54811953 on Build GRCh38
rs763318737 Chr.1: 54811885-54811885 on Build GRCh38
rs1022924490 Chr.1: 54812062-54812062 on Build GRCh38
rs368521655 Chr.1: 54811899-54811899 on Build GRCh38
rs960369699 Chr.1: 54811901-54811901 on Build GRCh38
rs150096503 Chr.1: 54811917-54811917 on Build GRCh38
rs367644584 Chr.1: 54811942-54811942 on Build GRCh38
rs982790976 Chr.1: 54811955-54811955 on Build GRCh38
rs200524485 Chr.1: 54811976-54811976 on Build GRCh38
rs759538247 Chr.1: 54811910-54811910 on Build GRCh38
rs780036457 Chr.1: 54812037-54812037 on Build GRCh38
rs767668197 Chr.1: 54812012-54812012 on Build GRCh38
rs61691036 Chr.1: 54811880-54811880 on Build GRCh38
rs980431218 Chr.1: 54811896-54811896 on Build GRCh38
rs373140470 Chr.1: 54811969-54811969 on Build GRCh38
rs147721272 Chr.1: 54811972-54811972 on Build GRCh38
rs780559977 Chr.1: 54811939-54811939 on Build GRCh38
rs749777216 Chr.1: 54812053-54812053 on Build GRCh38
rs765159171 Chr.1: 54811914-54811914 on Build GRCh38
rs369874938 Chr.1: 54811961-54811961 on Build GRCh38
rs1002422438 Chr.1: 54812064-54812064 on Build GRCh38
rs140694038 Chr.1: 54811887-54811887 on Build GRCh38
rs371945610 Chr.1: 54811916-54811916 on Build GRCh38
rs139677684 Chr.1: 54811881-54811881 on Build GRCh38
rs970067420 Chr.1: 54811882-54811882 on Build GRCh38
rs600499 Chr.1: 54811926-54811926 on Build GRCh38
rs756177298 Chr.1: 54812031-54812031 on Build GRCh38
rs777462327 Chr.1: 54811920-54811920 on Build GRCh38
rs375219574 Chr.1: 54811954-54811954 on Build GRCh38
rs182195719 Chr.1: 54811941-54811941 on Build GRCh38
rs867793576 Chr.1: 54811875-54811875 on Build GRCh38
rs781236008 Chr.1: 54811931-54811931 on Build GRCh38
rs749788935 Chr.1: 54811940-54811940 on Build GRCh38
rs760440349 Chr.1: 54811892-54811892 on Build GRCh38
rs750665751 Chr.1: 54812017-54812017 on Build GRCh38
rs760386820 Chr.1: 54811952-54811952 on Build GRCh38
rs755493995 Chr.1: 54812063-54812063 on Build GRCh38
rs751414286 Chr.1: 54811997-54811997 on Build GRCh38

View Full Product Details