Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 40057096-40057369 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BCOR
Entrez Gene ID
54880
Gene Name
BCL6 corepressor
Gene Aliases
ANOP2, MAA2, MCOPS2
UniGene
Hs.659681
Gene Chromosome Location
Chr.X: 40051246-40177390 on Build GRCh38
Transcripts
NM_001123383.1 NM_001123384.1 NM_001123385.1 NM_017745.5 XM_005272616.1 XM_005272618.3 XM_005272619.3 XM_005272620.3 XM_006724536.3 XM_011543929.2 XM_011543930.1 XM_011543931.2 XM_017029615.1 XM_017029616.1

SNP Detail

SNP ID Chromosome Location
rs748764772 Chr.X: 40057329-40057329 on Build GRCh38
rs745562627 Chr.X: 40057344-40057344 on Build GRCh38
rs374206405 Chr.X: 40057172-40057172 on Build GRCh38
rs770413249 Chr.X: 40057330-40057330 on Build GRCh38
rs138741225 Chr.X: 40057262-40057262 on Build GRCh38
rs368796810 Chr.X: 40057217-40057217 on Build GRCh38
rs754796420 Chr.X: 40057265-40057265 on Build GRCh38
rs764494664 Chr.X: 40057189-40057189 on Build GRCh38
rs770555045 Chr.X: 40057146-40057146 on Build GRCh38
rs761093745 Chr.X: 40057187-40057187 on Build GRCh38
rs199676230 Chr.X: 40057210-40057210 on Build GRCh38
rs375830184 Chr.X: 40057190-40057190 on Build GRCh38
rs750459436 Chr.X: 40057205-40057205 on Build GRCh38
rs778612759 Chr.X: 40057336-40057336 on Build GRCh38
rs765759611 Chr.X: 40057200-40057200 on Build GRCh38
rs781170619 Chr.X: 40057276-40057276 on Build GRCh38
rs756063203 Chr.X: 40057316-40057316 on Build GRCh38
rs758631631 Chr.X: 40057169-40057169 on Build GRCh38
rs777585092 Chr.X: 40057318-40057318 on Build GRCh38
rs766620661 Chr.X: 40057253-40057253 on Build GRCh38
rs375344311 Chr.X: 40057277-40057277 on Build GRCh38
rs372724488 Chr.X: 40057228-40057228 on Build GRCh38

View Full Product Details