Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 87509354-87509581 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ABCB1
Entrez Gene ID
5243
Gene Name
ATP binding cassette subfamily B member 1
Gene Aliases
ABC20, CD243, CLCS, GP170, MDR1, P-GP, PGY1
UniGene
Hs.489033
Gene Chromosome Location
Chr.7: 87503863-87713323 on Build GRCh38
Transcripts
NM_000927.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4488671 c.3364C>T p.L1122L Chr.7: 87509400-87509400 on Build GRCh38
227035 c.3355G>A p.E1119K Chr.7: 87509409-87509409 on Build GRCh38
4149387 c.3349T>A p.S1117T Chr.7: 87509415-87509415 on Build GRCh38
6568329 c.3298G>A p.E1100K Chr.7: 87509466-87509466 on Build GRCh38
4649141 c.3288T>C p.L1096L Chr.7: 87509476-87509476 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs972508453 Chr.7: 87509541-87509541 on Build GRCh38
rs371241634 Chr.7: 87509470-87509470 on Build GRCh38
rs766133333 Chr.7: 87509494-87509494 on Build GRCh38
rs185002393 Chr.7: 87509458-87509458 on Build GRCh38
rs28401807 Chr.7: 87509549-87509549 on Build GRCh38
rs201143383 Chr.7: 87509395-87509395 on Build GRCh38
rs747669868 Chr.7: 87509513-87509513 on Build GRCh38
rs1017877229 Chr.7: 87509401-87509401 on Build GRCh38
rs955026052 Chr.7: 87509552-87509552 on Build GRCh38
rs200182870 Chr.7: 87509503-87509503 on Build GRCh38
rs763091787 Chr.7: 87509379-87509379 on Build GRCh38
rs202215242 Chr.7: 87509402-87509402 on Build GRCh38
rs189804819 Chr.7: 87509504-87509504 on Build GRCh38
rs536000511 Chr.7: 87509416-87509416 on Build GRCh38
rs61040122 Chr.7: 87509531-87509531 on Build GRCh38
rs55852620 Chr.7: 87509444-87509444 on Build GRCh38
rs755677028 Chr.7: 87509530-87509530 on Build GRCh38
rs41309225 Chr.7: 87509469-87509469 on Build GRCh38
rs747650743 Chr.7: 87509424-87509424 on Build GRCh38
rs770289924 Chr.7: 87509435-87509435 on Build GRCh38
rs949343048 Chr.7: 87509542-87509542 on Build GRCh38
rs1014207158 Chr.7: 87509436-87509436 on Build GRCh38
rs909739867 Chr.7: 87509500-87509500 on Build GRCh38
rs780256270 Chr.7: 87509494-87509495 on Build GRCh38
rs779984514 Chr.7: 87509400-87509400 on Build GRCh38
rs199894992 Chr.7: 87509385-87509385 on Build GRCh38
rs148897157 Chr.7: 87509448-87509448 on Build GRCh38
rs746677023 Chr.7: 87509405-87509405 on Build GRCh38
rs780586109 Chr.7: 87509419-87509419 on Build GRCh38
rs749835902 Chr.7: 87509532-87509532 on Build GRCh38
rs377258612 Chr.7: 87509525-87509525 on Build GRCh38
rs749468893 Chr.7: 87509515-87509515 on Build GRCh38
rs372391015 Chr.7: 87509392-87509392 on Build GRCh38
rs199931681 Chr.7: 87509420-87509420 on Build GRCh38
rs201204429 Chr.7: 87509449-87509449 on Build GRCh38
rs758212180 Chr.7: 87509391-87509391 on Build GRCh38
rs774748307 Chr.7: 87509445-87509445 on Build GRCh38
rs201566913 Chr.7: 87509506-87509506 on Build GRCh38
rs758377034 Chr.7: 87509476-87509476 on Build GRCh38
rs773651038 Chr.7: 87509439-87509439 on Build GRCh38
rs35730308 Chr.7: 87509442-87509442 on Build GRCh38
rs750446497 Chr.7: 87509473-87509473 on Build GRCh38

View Full Product Details