Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 99424947-99425220 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATP5J2-PTCD1
Entrez Gene ID
100526740
Gene Name
ATP5J2-PTCD1 readthrough
Gene Aliases
-
UniGene
Hs.632313
Gene Chromosome Location
Chr.7: 99416739-99466201 on Build GRCh38
Transcripts
NM_001198879.1 NM_015545.3

Gene Symbol
PTCD1
Entrez Gene ID
26024
Gene Name
pentatricopeptide repeat domain 1
Gene Aliases
-
UniGene
Hs.632313
Gene Chromosome Location
Chr.7: 99416739-99438839 on Build GRCh38
Transcripts
NM_001198879.1 NM_015545.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3833599 c.1531G>C p.A511P Chr.7: 99425148-99425148 on Build GRCh38
4785537 c.1483G>A p.G495R Chr.7: 99425196-99425196 on Build GRCh38
4925896 c.1712T>C p.F571S Chr.7: 99424967-99424967 on Build GRCh38
6901904 c.1549G>A p.E517K Chr.7: 99424983-99424983 on Build GRCh38
6515297 c.1518G>T p.T506T Chr.7: 99425161-99425161 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs757091950 Chr.7: 99425087-99425087 on Build GRCh38
rs773504003 Chr.7: 99425037-99425037 on Build GRCh38
rs911202630 Chr.7: 99425110-99425110 on Build GRCh38
rs770517214 Chr.7: 99425079-99425084 on Build GRCh38
rs771263062 Chr.7: 99425145-99425145 on Build GRCh38
rs573252963 Chr.7: 99425161-99425161 on Build GRCh38
rs145450760 Chr.7: 99425076-99425076 on Build GRCh38
rs774730987 Chr.7: 99425146-99425146 on Build GRCh38
rs906905288 Chr.7: 99424993-99424993 on Build GRCh38
rs942946815 Chr.7: 99425125-99425125 on Build GRCh38
rs751269088 Chr.7: 99424975-99424975 on Build GRCh38
rs777645699 Chr.7: 99424998-99424998 on Build GRCh38
rs112219883 Chr.7: 99425105-99425105 on Build GRCh38
rs756153809 Chr.7: 99425199-99425199 on Build GRCh38
rs759256492 Chr.7: 99425049-99425049 on Build GRCh38
rs868292144 Chr.7: 99425100-99425100 on Build GRCh38
rs749833988 Chr.7: 99425142-99425142 on Build GRCh38
rs111662245 Chr.7: 99425162-99425162 on Build GRCh38
rs375649241 Chr.7: 99425200-99425200 on Build GRCh38
rs201657058 Chr.7: 99425112-99425112 on Build GRCh38
rs971289603 Chr.7: 99425189-99425189 on Build GRCh38
rs770336095 Chr.7: 99425035-99425035 on Build GRCh38
rs148440637 Chr.7: 99425129-99425129 on Build GRCh38
rs765081795 Chr.7: 99425186-99425186 on Build GRCh38
rs1017439389 Chr.7: 99425063-99425063 on Build GRCh38
rs369313527 Chr.7: 99425143-99425143 on Build GRCh38
rs753808492 Chr.7: 99425181-99425181 on Build GRCh38
rs556087930 Chr.7: 99425179-99425179 on Build GRCh38
rs373268743 Chr.7: 99425141-99425141 on Build GRCh38
rs747278474 Chr.7: 99425010-99425010 on Build GRCh38
rs759044184 Chr.7: 99425153-99425153 on Build GRCh38
rs756690997 Chr.7: 99425191-99425191 on Build GRCh38
rs752528450 Chr.7: 99424988-99424988 on Build GRCh38
rs778903791 Chr.7: 99425094-99425094 on Build GRCh38
rs751679542 Chr.7: 99425104-99425104 on Build GRCh38
rs750945895 Chr.7: 99425163-99425163 on Build GRCh38
rs377292526 Chr.7: 99425006-99425006 on Build GRCh38
rs35500147 Chr.7: 99425197-99425197 on Build GRCh38
rs192898483 Chr.7: 99425001-99425001 on Build GRCh38
rs916794538 Chr.7: 99425175-99425175 on Build GRCh38
rs114157830 Chr.7: 99425018-99425018 on Build GRCh38
rs920095829 Chr.7: 99425131-99425131 on Build GRCh38
rs775458723 Chr.7: 99424978-99424978 on Build GRCh38
rs200408854 Chr.7: 99425190-99425190 on Build GRCh38
rs748559296 Chr.7: 99425034-99425034 on Build GRCh38
rs201202678 Chr.7: 99425050-99425050 on Build GRCh38
rs535281099 Chr.7: 99425060-99425060 on Build GRCh38
rs773961536 Chr.7: 99425047-99425047 on Build GRCh38
rs756107991 Chr.7: 99424990-99424990 on Build GRCh38
rs761867510 Chr.7: 99425185-99425185 on Build GRCh38
rs373000640 Chr.7: 99425196-99425196 on Build GRCh38
rs771351214 Chr.7: 99425044-99425044 on Build GRCh38
rs768600193 Chr.7: 99425011-99425011 on Build GRCh38
rs763289798 Chr.7: 99425043-99425043 on Build GRCh38
rs140529605 Chr.7: 99425140-99425140 on Build GRCh38
rs369540728 Chr.7: 99425077-99425077 on Build GRCh38
rs774639475 Chr.7: 99424977-99424977 on Build GRCh38

View Full Product Details