Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 58131573-58131800 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C14orf37
Entrez Gene ID
145407
Gene Name
chromosome 14 open reading frame 37
Gene Aliases
c14_5376
UniGene
Hs.26766
Gene Chromosome Location
Chr.14: 57993536-58152305 on Build GRCh38
Transcripts
NM_001001872.3 NM_001320173.1 XM_011536471.2 XM_017021029.1 XM_017021030.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
159492 c.1708G>A p.E570K Chr.14: 58131635-58131635 on Build GRCh38
7358063 c.1687C>A p.P563T Chr.14: 58131656-58131656 on Build GRCh38
6477031 c.1664C>T p.P555L Chr.14: 58131679-58131679 on Build GRCh38
256443 c.1669C>A p.L557M Chr.14: 58131674-58131674 on Build GRCh38
6333167 c.1622-4G>T p.? Chr.14: 58131725-58131725 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs893868106 Chr.14: 58131753-58131753 on Build GRCh38
rs772324448 Chr.14: 58131687-58131687 on Build GRCh38
rs774817631 Chr.14: 58131618-58131618 on Build GRCh38
rs745388735 Chr.14: 58131749-58131749 on Build GRCh38
rs199514609 Chr.14: 58131707-58131707 on Build GRCh38
rs147141968 Chr.14: 58131690-58131692 on Build GRCh38
rs772692558 Chr.14: 58131632-58131632 on Build GRCh38
rs780704027 Chr.14: 58131665-58131665 on Build GRCh38
rs904827589 Chr.14: 58131744-58131744 on Build GRCh38
rs45570237 Chr.14: 58131719-58131719 on Build GRCh38
rs775703964 Chr.14: 58131635-58131635 on Build GRCh38
rs751504037 Chr.14: 58131658-58131658 on Build GRCh38
rs144281427 Chr.14: 58131655-58131655 on Build GRCh38
rs45465593 Chr.14: 58131639-58131639 on Build GRCh38
rs747814311 Chr.14: 58131599-58131599 on Build GRCh38
rs746306058 Chr.14: 58131679-58131679 on Build GRCh38
rs886951463 Chr.14: 58131657-58131657 on Build GRCh38
rs754062638 Chr.14: 58131646-58131646 on Build GRCh38
rs762742883 Chr.14: 58131741-58131741 on Build GRCh38
rs372804317 Chr.14: 58131643-58131643 on Build GRCh38
rs754946059 Chr.14: 58131767-58131767 on Build GRCh38
rs868130333 Chr.14: 58131687-58131689 on Build GRCh38
rs776893510 Chr.14: 58131718-58131718 on Build GRCh38
rs754026877 Chr.14: 58131676-58131676 on Build GRCh38
rs779132277 Chr.14: 58131677-58131677 on Build GRCh38
rs991596361 Chr.14: 58131606-58131606 on Build GRCh38
rs776080070 Chr.14: 58131702-58131702 on Build GRCh38
rs571704933 Chr.14: 58131647-58131647 on Build GRCh38
rs764912913 Chr.14: 58131651-58131651 on Build GRCh38
rs771159119 Chr.14: 58131615-58131615 on Build GRCh38
rs758259560 Chr.14: 58131653-58131653 on Build GRCh38
rs149864826 Chr.14: 58131598-58131598 on Build GRCh38
rs754429032 Chr.14: 58131659-58131659 on Build GRCh38
rs747722762 Chr.14: 58131667-58131667 on Build GRCh38
rs200555210 Chr.14: 58131709-58131709 on Build GRCh38
rs35930281 Chr.14: 58131669-58131670 on Build GRCh38
rs750285376 Chr.14: 58131652-58131652 on Build GRCh38
rs759937754 Chr.14: 58131622-58131622 on Build GRCh38
rs772879260 Chr.14: 58131722-58131722 on Build GRCh38
rs182305719 Chr.14: 58131747-58131747 on Build GRCh38
rs751451261 Chr.14: 58131757-58131757 on Build GRCh38
rs199639523 Chr.14: 58131623-58131623 on Build GRCh38
rs201126356 Chr.14: 58131711-58131711 on Build GRCh38
rs145807752 Chr.14: 58131619-58131619 on Build GRCh38
rs62005364 Chr.14: 58131769-58131769 on Build GRCh38
rs973591724 Chr.14: 58131629-58131629 on Build GRCh38

View Full Product Details