Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 75166767-75167000 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZFP1
Entrez Gene ID
162239
Gene Name
ZFP1 zinc finger protein
Gene Aliases
ZNF475
UniGene
Hs.388813
Gene Chromosome Location
Chr.16: 75126285-75172234 on Build GRCh38
Transcripts
NM_001318469.1 NM_001318471.1 NM_001318472.1 NM_001318473.1 NM_001318474.1 NM_001318475.1 NM_001318476.1 NM_153688.3 XM_011522921.2 XM_017022983.1 XM_017022984.1 XM_017022985.1 XM_017022986.1 XM_017022987.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7192270 c.16-2A>T p.? Chr.16: 75166768-75166768 on Build GRCh38
6432480 c.142G>A p.E48K Chr.16: 75166896-75166896 on Build GRCh38
2691094 c.63A>G p.E21E Chr.16: 75166817-75166817 on Build GRCh38
4801182 c.33G>T p.T11T Chr.16: 75166787-75166787 on Build GRCh38
1379779 c.51T>C p.F17F Chr.16: 75166805-75166805 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs148482862 Chr.16: 75166925-75166925 on Build GRCh38
rs375285545 Chr.16: 75166914-75166914 on Build GRCh38
rs781458982 Chr.16: 75166847-75166847 on Build GRCh38
rs905830498 Chr.16: 75166913-75166913 on Build GRCh38
rs752630529 Chr.16: 75166919-75166919 on Build GRCh38
rs954372849 Chr.16: 75166964-75166964 on Build GRCh38
rs778062146 Chr.16: 75166958-75166958 on Build GRCh38
rs777476771 Chr.16: 75166904-75166904 on Build GRCh38
rs753435956 Chr.16: 75166941-75166941 on Build GRCh38
rs368243920 Chr.16: 75166920-75166920 on Build GRCh38
rs767168661 Chr.16: 75166812-75166812 on Build GRCh38
rs754858967 Chr.16: 75166835-75166835 on Build GRCh38
rs367856248 Chr.16: 75166929-75166929 on Build GRCh38
rs769320379 Chr.16: 75166799-75166799 on Build GRCh38
rs773664855 Chr.16: 75166795-75166795 on Build GRCh38
rs757302319 Chr.16: 75166968-75166968 on Build GRCh38
rs569308046 Chr.16: 75166932-75166932 on Build GRCh38
rs762134697 Chr.16: 75166802-75166802 on Build GRCh38
rs747421736 Chr.16: 75166962-75166962 on Build GRCh38
rs752502333 Chr.16: 75166839-75166839 on Build GRCh38
rs368944427 Chr.16: 75166877-75166877 on Build GRCh38
rs765134367 Chr.16: 75166838-75166838 on Build GRCh38
rs776095270 Chr.16: 75166912-75166912 on Build GRCh38
rs1049075771 Chr.16: 75166966-75166966 on Build GRCh38
rs755081449 Chr.16: 75166945-75166945 on Build GRCh38
rs968152197 Chr.16: 75166856-75166856 on Build GRCh38
rs763079554 Chr.16: 75166801-75166801 on Build GRCh38
rs756358635 Chr.16: 75166933-75166933 on Build GRCh38
rs767991481 Chr.16: 75166827-75166828 on Build GRCh38
rs779462924 Chr.16: 75166950-75166950 on Build GRCh38
rs1016432946 Chr.16: 75166953-75166953 on Build GRCh38
rs765214265 Chr.16: 75166917-75166917 on Build GRCh38
rs748543875 Chr.16: 75166901-75166901 on Build GRCh38
rs779330114 Chr.16: 75166893-75166893 on Build GRCh38
rs751742960 Chr.16: 75166841-75166841 on Build GRCh38
rs201833464 Chr.16: 75166793-75166793 on Build GRCh38
rs144579719 Chr.16: 75166909-75166909 on Build GRCh38
rs765808592 Chr.16: 75166829-75166829 on Build GRCh38
rs760279673 Chr.16: 75166905-75166905 on Build GRCh38
rs753126272 Chr.16: 75166877-75166878 on Build GRCh38
rs778140504 Chr.16: 75166930-75166934 on Build GRCh38
rs748546982 Chr.16: 75166951-75166951 on Build GRCh38
rs756413734 Chr.16: 75166850-75166850 on Build GRCh38
rs753939292 Chr.16: 75166940-75166940 on Build GRCh38
rs755069693 Chr.16: 75166866-75166866 on Build GRCh38
rs963919121 Chr.16: 75166955-75166955 on Build GRCh38
rs756458132 Chr.16: 75166917-75166918 on Build GRCh38
rs867484972 Chr.16: 75166911-75166911 on Build GRCh38
rs537167395 Chr.16: 75166956-75166956 on Build GRCh38
rs201225610 Chr.16: 75166840-75166840 on Build GRCh38
rs144244672 Chr.16: 75166821-75166821 on Build GRCh38
rs909404057 Chr.16: 75166797-75166797 on Build GRCh38
rs549740432 Chr.16: 75166928-75166928 on Build GRCh38
rs371827111 Chr.16: 75166934-75166934 on Build GRCh38
rs772807692 Chr.16: 75166817-75166817 on Build GRCh38
rs746026233 Chr.16: 75166848-75166848 on Build GRCh38
rs774731795 Chr.16: 75166800-75166800 on Build GRCh38
rs963609627 Chr.16: 75166805-75166805 on Build GRCh38
rs780246252 Chr.16: 75166855-75166855 on Build GRCh38

View Full Product Details