Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 99970473-99970700 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZIC5
Entrez Gene ID
85416
Gene Name
Zic family member 5
Gene Aliases
-
UniGene
Hs.508570
Gene Chromosome Location
Chr.13: 99963021-99971924 on Build GRCh38
Transcripts
NM_033132.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4147541 c.1194C>A p.C398* Chr.13: 99970482-99970482 on Build GRCh38
4958451 c.1125C>A p.P375P Chr.13: 99970551-99970551 on Build GRCh38
7273600 c.1077G>A p.A359A Chr.13: 99970599-99970599 on Build GRCh38
6827264 c.1165C>T p.R389W Chr.13: 99970511-99970511 on Build GRCh38
6052881 c.977G>A p.G326D Chr.13: 99970699-99970699 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs770449936 Chr.13: 99970567-99970567 on Build GRCh38
rs905616806 Chr.13: 99970573-99970573 on Build GRCh38
rs769238260 Chr.13: 99970510-99970510 on Build GRCh38
rs776567415 Chr.13: 99970594-99970605 on Build GRCh38
rs779482891 Chr.13: 99970557-99970558 on Build GRCh38
rs763585410 Chr.13: 99970497-99970497 on Build GRCh38
rs766633692 Chr.13: 99970629-99970629 on Build GRCh38
rs773386049 Chr.13: 99970581-99970581 on Build GRCh38
rs746677608 Chr.13: 99970563-99970563 on Build GRCh38
rs532287066 Chr.13: 99970583-99970584 on Build GRCh38
rs912633798 Chr.13: 99970624-99970624 on Build GRCh38
rs769398079 Chr.13: 99970571-99970571 on Build GRCh38
rs370372534 Chr.13: 99970537-99970537 on Build GRCh38
rs963617173 Chr.13: 99970585-99970585 on Build GRCh38
rs752692151 Chr.13: 99970649-99970649 on Build GRCh38
rs746163974 Chr.13: 99970579-99970596 on Build GRCh38
rs760991645 Chr.13: 99970590-99970590 on Build GRCh38
rs750717552 Chr.13: 99970527-99970527 on Build GRCh38
rs774978018 Chr.13: 99970514-99970514 on Build GRCh38
rs758758105 Chr.13: 99970588-99970605 on Build GRCh38
rs762280576 Chr.13: 99970516-99970516 on Build GRCh38
rs866031645 Chr.13: 99970520-99970520 on Build GRCh38
rs755023690 Chr.13: 99970539-99970539 on Build GRCh38
rs776213644 Chr.13: 99970496-99970496 on Build GRCh38
rs927426122 Chr.13: 99970667-99970667 on Build GRCh38
rs937510913 Chr.13: 99970670-99970670 on Build GRCh38
rs778858860 Chr.13: 99970541-99970541 on Build GRCh38
rs768818644 Chr.13: 99970591-99970605 on Build GRCh38
rs745419960 Chr.13: 99970568-99970568 on Build GRCh38
rs956520549 Chr.13: 99970565-99970588 on Build GRCh38
rs954172816 Chr.13: 99970596-99970596 on Build GRCh38
rs760754333 Chr.13: 99970530-99970530 on Build GRCh38
rs762216116 Chr.13: 99970579-99970579 on Build GRCh38
rs965409885 Chr.13: 99970637-99970637 on Build GRCh38
rs752569231 Chr.13: 99970550-99970550 on Build GRCh38
rs765284967 Chr.13: 99970648-99970648 on Build GRCh38
rs772502232 Chr.13: 99970580-99970580 on Build GRCh38
rs913484014 Chr.13: 99970526-99970526 on Build GRCh38
rs186203798 Chr.13: 99970638-99970638 on Build GRCh38
rs759676532 Chr.13: 99970644-99970644 on Build GRCh38
rs1029202774 Chr.13: 99970595-99970595 on Build GRCh38
rs947616255 Chr.13: 99970557-99970557 on Build GRCh38
rs775066063 Chr.13: 99970578-99970578 on Build GRCh38
rs566928909 Chr.13: 99970561-99970561 on Build GRCh38
rs376208865 Chr.13: 99970495-99970495 on Build GRCh38
rs780655431 Chr.13: 99970584-99970586 on Build GRCh38
rs766455576 Chr.13: 99970531-99970531 on Build GRCh38
rs758268448 Chr.13: 99970554-99970554 on Build GRCh38
rs985978093 Chr.13: 99970603-99970603 on Build GRCh38
rs546848236 Chr.13: 99970675-99970675 on Build GRCh38
rs913496978 Chr.13: 99970646-99970646 on Build GRCh38
rs767771682 Chr.13: 99970519-99970519 on Build GRCh38

View Full Product Details