Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 65591606-65591849 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TRIM23
Entrez Gene ID
373
Gene Name
tripartite motif containing 23
Gene Aliases
ARD1, ARFD1, RNF46
UniGene
Hs.792
Gene Chromosome Location
Chr.5: 65589680-65624360 on Build GRCh38
Transcripts
NM_001656.3 NM_033227.2 NM_033228.2 XM_005248497.3 XM_017009444.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4529912 c.1651G>A p.G551S Chr.5: 65591843-65591843 on Build GRCh38
4864858 c.1645C>T p.R549* Chr.5: 65591849-65591849 on Build GRCh38
3356043 c.1646G>A p.R549Q Chr.5: 65591848-65591848 on Build GRCh38
2150966 c.1655T>C p.M552T Chr.5: 65591839-65591839 on Build GRCh38
1069357 c.1688G>A p.R563Q Chr.5: 65591806-65591806 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs760151704 Chr.5: 65591751-65591752 on Build GRCh38
rs35400101 Chr.5: 65591646-65591647 on Build GRCh38
rs965021949 Chr.5: 65591692-65591692 on Build GRCh38
rs879059096 Chr.5: 65591646-65591647 on Build GRCh38
rs1021757371 Chr.5: 65591645-65591645 on Build GRCh38
rs546107130 Chr.5: 65591749-65591751 on Build GRCh38
rs1057028280 Chr.5: 65591822-65591822 on Build GRCh38
rs143394952 Chr.5: 65591817-65591817 on Build GRCh38
rs1007133400 Chr.5: 65591649-65591649 on Build GRCh38
rs969417225 Chr.5: 65591648-65591648 on Build GRCh38
rs10644434 Chr.5: 65591645-65591646 on Build GRCh38
rs61748229 Chr.5: 65591787-65591787 on Build GRCh38
rs962238139 Chr.5: 65591659-65591660 on Build GRCh38
rs768458224 Chr.5: 65591806-65591806 on Build GRCh38
rs554972458 Chr.5: 65591654-65591654 on Build GRCh38
rs757128503 Chr.5: 65591760-65591760 on Build GRCh38
rs746829642 Chr.5: 65591751-65591751 on Build GRCh38
rs1044971019 Chr.5: 65591783-65591783 on Build GRCh38
rs909712980 Chr.5: 65591769-65591769 on Build GRCh38
rs66680365 Chr.5: 65591666-65591667 on Build GRCh38
rs745886720 Chr.5: 65591777-65591777 on Build GRCh38
rs574915364 Chr.5: 65591656-65591656 on Build GRCh38
rs972043165 Chr.5: 65591671-65591671 on Build GRCh38
rs755268241 Chr.5: 65591718-65591718 on Build GRCh38
rs918352630 Chr.5: 65591745-65591745 on Build GRCh38
rs933024511 Chr.5: 65591730-65591731 on Build GRCh38
rs909054225 Chr.5: 65591647-65591647 on Build GRCh38
rs777503087 Chr.5: 65591749-65591749 on Build GRCh38
rs543719525 Chr.5: 65591674-65591674 on Build GRCh38
rs775117910 Chr.5: 65591790-65591790 on Build GRCh38
rs748865849 Chr.5: 65591792-65591792 on Build GRCh38
rs35769160 Chr.5: 65591775-65591775 on Build GRCh38

View Full Product Details