Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 743085-743320 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
YES1
Entrez Gene ID
7525
Gene Name
YES proto-oncogene 1, Src family tyrosine kinase
Gene Aliases
HsT441, P61-YES, Yes, c-yes
UniGene
Hs.194148
Gene Chromosome Location
Chr.18: 721592-812326 on Build GRCh38
Transcripts
NM_005433.3 XM_005258139.3 XM_017025960.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4440509 c.832C>T p.R278* Chr.18: 743308-743308 on Build GRCh38
7140662 c.833G>T p.R278L Chr.18: 743307-743307 on Build GRCh38
6824373 c.833G>A p.R278Q Chr.18: 743307-743307 on Build GRCh38
6824376 c.831G>A p.L277L Chr.18: 743309-743309 on Build GRCh38
3959401 c.825A>T p.E275D Chr.18: 743315-743315 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs896250821 Chr.18: 743126-743126 on Build GRCh38
rs748787911 Chr.18: 743126-743135 on Build GRCh38
rs913245471 Chr.18: 743134-743134 on Build GRCh38
rs779712187 Chr.18: 743210-743210 on Build GRCh38
rs750062336 Chr.18: 743136-743136 on Build GRCh38
rs199903309 Chr.18: 743286-743286 on Build GRCh38
rs763308490 Chr.18: 743124-743124 on Build GRCh38
rs753156030 Chr.18: 743119-743119 on Build GRCh38
rs865963468 Chr.18: 743222-743222 on Build GRCh38
rs747200359 Chr.18: 743263-743263 on Build GRCh38
rs536950970 Chr.18: 743228-743228 on Build GRCh38
rs753298023 Chr.18: 743215-743215 on Build GRCh38
rs200885771 Chr.18: 743250-743250 on Build GRCh38
rs763427480 Chr.18: 743293-743293 on Build GRCh38
rs368845120 Chr.18: 743273-743273 on Build GRCh38
rs755805607 Chr.18: 743141-743141 on Build GRCh38
rs778845997 Chr.18: 743235-743235 on Build GRCh38
rs772604774 Chr.18: 743138-743139 on Build GRCh38
rs775958102 Chr.18: 743145-743152 on Build GRCh38
rs555212897 Chr.18: 743240-743240 on Build GRCh38
rs777396959 Chr.18: 743122-743123 on Build GRCh38
rs897955047 Chr.18: 743198-743198 on Build GRCh38
rs777763921 Chr.18: 743252-743252 on Build GRCh38
rs1024484217 Chr.18: 743167-743167 on Build GRCh38
rs759600003 Chr.18: 743275-743275 on Build GRCh38
rs764433044 Chr.18: 743127-743127 on Build GRCh38
rs771891606 Chr.18: 743244-743244 on Build GRCh38
rs35126906 Chr.18: 743295-743295 on Build GRCh38
rs776058735 Chr.18: 743287-743287 on Build GRCh38
rs1028355291 Chr.18: 743231-743231 on Build GRCh38
rs571331504 Chr.18: 743123-743123 on Build GRCh38
rs981375945 Chr.18: 743179-743179 on Build GRCh38
rs945993600 Chr.18: 743151-743151 on Build GRCh38
rs188857712 Chr.18: 743272-743272 on Build GRCh38
rs769766379 Chr.18: 743276-743276 on Build GRCh38
rs3786347 Chr.18: 743226-743226 on Build GRCh38

View Full Product Details