Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 61393146-61393419 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TMEM216
Entrez Gene ID
51259
Gene Name
transmembrane protein 216
Gene Aliases
HSPC244
UniGene
Hs.26745
Gene Chromosome Location
Chr.11: 61391687-61398863 on Build GRCh38
Transcripts
NM_001173990.2 NM_001173991.2 NM_016499.5 XM_005274039.3 XM_017017882.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5799263 c.78G>A p.L26L Chr.11: 61393274-61393274 on Build GRCh38
5999729 c.68T>C p.L23P Chr.11: 61393264-61393264 on Build GRCh38
5799264 c.78G>A p.L26L Chr.11: 61393274-61393274 on Build GRCh38
5999728 c.68T>C p.L23P Chr.11: 61393264-61393264 on Build GRCh38
1161002 c.85T>A p.W29R Chr.11: 61393281-61393281 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs779378169 Chr.11: 61393359-61393359 on Build GRCh38
rs996913515 Chr.11: 61393205-61393205 on Build GRCh38
rs1045541681 Chr.11: 61393197-61393197 on Build GRCh38
rs141905811 Chr.11: 61393336-61393336 on Build GRCh38
rs528271337 Chr.11: 61393236-61393236 on Build GRCh38
rs568253718 Chr.11: 61393309-61393309 on Build GRCh38
rs144650416 Chr.11: 61393199-61393199 on Build GRCh38
rs376314259 Chr.11: 61393233-61393233 on Build GRCh38
rs920229849 Chr.11: 61393348-61393348 on Build GRCh38
rs189765321 Chr.11: 61393297-61393297 on Build GRCh38
rs1057517528 Chr.11: 61393229-61393229 on Build GRCh38
rs548233683 Chr.11: 61393247-61393247 on Build GRCh38
rs769285695 Chr.11: 61393253-61393253 on Build GRCh38
rs997518388 Chr.11: 61393342-61393342 on Build GRCh38
rs962913000 Chr.11: 61393299-61393299 on Build GRCh38
rs764098583 Chr.11: 61393199-61393200 on Build GRCh38
rs532474996 Chr.11: 61393174-61393174 on Build GRCh38
rs934536093 Chr.11: 61393212-61393212 on Build GRCh38
rs751485492 Chr.11: 61393353-61393355 on Build GRCh38
rs147953784 Chr.11: 61393214-61393214 on Build GRCh38
rs955708363 Chr.11: 61393390-61393390 on Build GRCh38
rs758077979 Chr.11: 61393302-61393302 on Build GRCh38
rs900061092 Chr.11: 61393319-61393319 on Build GRCh38
rs550981048 Chr.11: 61393365-61393365 on Build GRCh38
rs948665763 Chr.11: 61393191-61393191 on Build GRCh38
rs563417135 Chr.11: 61393172-61393172 on Build GRCh38

View Full Product Details