Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 117433283-117433524 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DSCAML1
Entrez Gene ID
57453
Gene Name
DS cell adhesion molecule like 1
Gene Aliases
DSCAM2
UniGene
Hs.659513
Gene Chromosome Location
Chr.11: 117427772-117797261 on Build GRCh38
Transcripts
NM_020693.3 XM_011542917.1 XM_011542918.1 XM_011542919.2 XM_011542920.2 XM_011542921.2 XM_011542924.2 XM_011542925.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
380847 c.5074G>C p.E1692Q Chr.11: 117433454-117433454 on Build GRCh38
5507640 c.5057-4G>T p.? Chr.11: 117433475-117433475 on Build GRCh38
5626415 c.5079G>T p.M1693I Chr.11: 117433449-117433449 on Build GRCh38
5371732 c.5058T>G p.D1686E Chr.11: 117433470-117433470 on Build GRCh38
3443801 c.5064G>A p.K1688K Chr.11: 117433464-117433464 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs548056744 Chr.11: 117433326-117433326 on Build GRCh38
rs754586952 Chr.11: 117433409-117433409 on Build GRCh38
rs1012254746 Chr.11: 117433422-117433422 on Build GRCh38
rs969536085 Chr.11: 117433318-117433318 on Build GRCh38
rs780846692 Chr.11: 117433417-117433417 on Build GRCh38
rs761712926 Chr.11: 117433303-117433303 on Build GRCh38
rs759762226 Chr.11: 117433301-117433301 on Build GRCh38
rs902335825 Chr.11: 117433367-117433367 on Build GRCh38
rs771343543 Chr.11: 117433482-117433482 on Build GRCh38
rs960752421 Chr.11: 117433412-117433412 on Build GRCh38
rs757423317 Chr.11: 117433431-117433432 on Build GRCh38
rs748491153 Chr.11: 117433423-117433423 on Build GRCh38
rs972279702 Chr.11: 117433432-117433432 on Build GRCh38
rs371233425 Chr.11: 117433370-117433370 on Build GRCh38
rs375428721 Chr.11: 117433430-117433430 on Build GRCh38
rs369988663 Chr.11: 117433304-117433304 on Build GRCh38
rs181236117 Chr.11: 117433364-117433364 on Build GRCh38
rs779480217 Chr.11: 117433395-117433395 on Build GRCh38
rs199872880 Chr.11: 117433311-117433311 on Build GRCh38
rs753634949 Chr.11: 117433302-117433302 on Build GRCh38
rs747434149 Chr.11: 117433406-117433406 on Build GRCh38
rs963546244 Chr.11: 117433387-117433387 on Build GRCh38
rs757832353 Chr.11: 117433307-117433307 on Build GRCh38
rs1016804484 Chr.11: 117433380-117433380 on Build GRCh38
rs35399839 Chr.11: 117433458-117433458 on Build GRCh38
rs1011237838 Chr.11: 117433328-117433328 on Build GRCh38
rs2242133 Chr.11: 117433480-117433480 on Build GRCh38
rs750440853 Chr.11: 117433305-117433305 on Build GRCh38
rs754153890 Chr.11: 117433407-117433408 on Build GRCh38
rs1035119932 Chr.11: 117433396-117433396 on Build GRCh38
rs751075167 Chr.11: 117433399-117433399 on Build GRCh38
rs1043601989 Chr.11: 117433348-117433348 on Build GRCh38

View Full Product Details