Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 7878949-7879222 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BMP6
Entrez Gene ID
654
Gene Name
bone morphogenetic protein 6
Gene Aliases
VGR, VGR1
UniGene
Hs.285671
Gene Chromosome Location
Chr.6: 7726778-7881728 on Build GRCh38
Transcripts
NM_001718.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
274141 c.1227A>C p.K409N Chr.6: 7879096-7879096 on Build GRCh38
6296357 c.1235G>T p.C412F Chr.6: 7879104-7879104 on Build GRCh38
6611968 c.1245T>C p.H415H Chr.6: 7879114-7879114 on Build GRCh38
3875660 c.1232C>T p.A411V Chr.6: 7879101-7879101 on Build GRCh38
1081822 c.1205-1G>T p.? Chr.6: 7879073-7879073 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs985039102 Chr.6: 7879020-7879020 on Build GRCh38
rs944617183 Chr.6: 7879064-7879064 on Build GRCh38
rs528564381 Chr.6: 7879002-7879003 on Build GRCh38
rs867099853 Chr.6: 7879012-7879012 on Build GRCh38
rs528902772 Chr.6: 7879083-7879083 on Build GRCh38
rs563044779 Chr.6: 7879159-7879159 on Build GRCh38
rs771233974 Chr.6: 7879051-7879051 on Build GRCh38
rs952021088 Chr.6: 7879016-7879016 on Build GRCh38
rs759662999 Chr.6: 7879076-7879076 on Build GRCh38
rs564912073 Chr.6: 7879167-7879167 on Build GRCh38
rs749928567 Chr.6: 7879168-7879168 on Build GRCh38
rs150962959 Chr.6: 7879013-7879013 on Build GRCh38
rs756925897 Chr.6: 7879125-7879125 on Build GRCh38
rs770040611 Chr.6: 7879037-7879037 on Build GRCh38
rs774681507 Chr.6: 7879169-7879169 on Build GRCh38
rs763089803 Chr.6: 7879050-7879050 on Build GRCh38
rs372724956 Chr.6: 7879160-7879160 on Build GRCh38
rs773566752 Chr.6: 7879040-7879040 on Build GRCh38
rs267172 Chr.6: 7879174-7879174 on Build GRCh38
rs778029814 Chr.6: 7879164-7879164 on Build GRCh38
rs752566872 Chr.6: 7879096-7879096 on Build GRCh38
rs760605331 Chr.6: 7879105-7879105 on Build GRCh38
rs746009850 Chr.6: 7879170-7879170 on Build GRCh38
rs776339955 Chr.6: 7879191-7879191 on Build GRCh38
rs750464456 Chr.6: 7879024-7879025 on Build GRCh38
rs764848513 Chr.6: 7879141-7879141 on Build GRCh38
rs370689056 Chr.6: 7879182-7879182 on Build GRCh38
rs376084684 Chr.6: 7879155-7879155 on Build GRCh38
rs781643305 Chr.6: 7879030-7879030 on Build GRCh38
rs780193172 Chr.6: 7879062-7879064 on Build GRCh38
rs559544299 Chr.6: 7878981-7878981 on Build GRCh38
rs772294681 Chr.6: 7879171-7879171 on Build GRCh38
rs372511357 Chr.6: 7879153-7879153 on Build GRCh38
rs748615416 Chr.6: 7879035-7879035 on Build GRCh38
rs369020252 Chr.6: 7879121-7879121 on Build GRCh38
rs763804588 Chr.6: 7879107-7879107 on Build GRCh38
rs1040280629 Chr.6: 7879117-7879117 on Build GRCh38
rs760565166 Chr.6: 7879176-7879176 on Build GRCh38
rs774591705 Chr.6: 7879075-7879075 on Build GRCh38
rs201017875 Chr.6: 7879089-7879089 on Build GRCh38
rs550970857 Chr.6: 7879025-7879040 on Build GRCh38
rs749977284 Chr.6: 7879148-7879148 on Build GRCh38
rs140759179 Chr.6: 7879165-7879165 on Build GRCh38

View Full Product Details