Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 30757437-30757702 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC189
Entrez Gene ID
90835
Gene Name
coiled-coil domain containing 189
Gene Aliases
C16orf93
UniGene
Hs.724010
Gene Chromosome Location
Chr.16: 30757226-30762277 on Build GRCh38
Transcripts
NM_001014979.2 XM_011545976.1 XM_011545978.1 XM_017023852.1 XM_017023853.1 XM_017023854.1 XM_017023855.1 XM_017023856.1 XM_017023857.1 XM_017023858.1 XM_017023859.1 XM_017023860.1 XM_017023861.1 XM_017023862.1 NM_000294.2 NM_001172432.1

Gene Symbol
PHKG2
Entrez Gene ID
5261
Gene Name
phosphorylase kinase catalytic subunit gamma 2
Gene Aliases
GSD9C
UniGene
Hs.65735
Gene Chromosome Location
Chr.16: 30748299-30761176 on Build GRCh38
Transcripts
NM_001014979.2 XM_011545976.1 XM_011545978.1 XM_017023852.1 XM_017023853.1 XM_017023854.1 XM_017023855.1 XM_017023856.1 XM_017023857.1 XM_017023858.1 XM_017023859.1 XM_017023860.1 XM_017023861.1 XM_017023862.1 NM_000294.2 NM_001172432.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5643529 c.871G>C p.E291Q Chr.16: 30757601-30757601 on Build GRCh38
4060231 c.801C>T p.S267S Chr.16: 30757560-30757560 on Build GRCh38
4060230 c.912C>T p.S304S Chr.16: 30757560-30757560 on Build GRCh38
4060233 c.788G>A p.R263Q Chr.16: 30757573-30757573 on Build GRCh38
4060232 c.899G>A p.R300Q Chr.16: 30757573-30757573 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs926575163 Chr.16: 30757553-30757553 on Build GRCh38
rs973232049 Chr.16: 30757583-30757583 on Build GRCh38
rs776921895 Chr.16: 30757523-30757523 on Build GRCh38
rs746708732 Chr.16: 30757471-30757471 on Build GRCh38
rs765937812 Chr.16: 30757592-30757592 on Build GRCh38
rs182267845 Chr.16: 30757574-30757574 on Build GRCh38
rs186003632 Chr.16: 30757644-30757644 on Build GRCh38
rs982058785 Chr.16: 30757551-30757551 on Build GRCh38
rs958132881 Chr.16: 30757499-30757499 on Build GRCh38
rs57192967 Chr.16: 30757680-30757680 on Build GRCh38
rs761692751 Chr.16: 30757645-30757645 on Build GRCh38
rs771223442 Chr.16: 30757503-30757503 on Build GRCh38
rs1034759971 Chr.16: 30757538-30757538 on Build GRCh38
rs754416154 Chr.16: 30757541-30757543 on Build GRCh38
rs757905209 Chr.16: 30757587-30757587 on Build GRCh38
rs988618534 Chr.16: 30757504-30757504 on Build GRCh38
rs138413324 Chr.16: 30757573-30757573 on Build GRCh38
rs199800099 Chr.16: 30757659-30757659 on Build GRCh38
rs748023608 Chr.16: 30757605-30757606 on Build GRCh38
rs959149640 Chr.16: 30757544-30757544 on Build GRCh38
rs775575214 Chr.16: 30757559-30757559 on Build GRCh38
rs756537669 Chr.16: 30757671-30757671 on Build GRCh38
rs778258937 Chr.16: 30757491-30757491 on Build GRCh38
rs572438853 Chr.16: 30757522-30757522 on Build GRCh38
rs768683366 Chr.16: 30757643-30757643 on Build GRCh38
rs1015599008 Chr.16: 30757586-30757586 on Build GRCh38
rs373334034 Chr.16: 30757637-30757637 on Build GRCh38
rs201418865 Chr.16: 30757636-30757636 on Build GRCh38
rs780805572 Chr.16: 30757569-30757592 on Build GRCh38
rs757370237 Chr.16: 30757634-30757634 on Build GRCh38
rs920413584 Chr.16: 30757589-30757589 on Build GRCh38
rs951819094 Chr.16: 30757569-30757569 on Build GRCh38
rs200334105 Chr.16: 30757562-30757562 on Build GRCh38
rs764727003 Chr.16: 30757652-30757652 on Build GRCh38
rs749449323 Chr.16: 30757492-30757492 on Build GRCh38
rs772079956 Chr.16: 30757552-30757552 on Build GRCh38
rs760762883 Chr.16: 30757560-30757560 on Build GRCh38
rs1042716708 Chr.16: 30757484-30757484 on Build GRCh38
rs765849404 Chr.16: 30757662-30757662 on Build GRCh38
rs753185535 Chr.16: 30757601-30757601 on Build GRCh38
rs1003387879 Chr.16: 30757525-30757525 on Build GRCh38
rs764975253 Chr.16: 30757582-30757582 on Build GRCh38
rs748444172 Chr.16: 30757463-30757463 on Build GRCh38
rs777968026 Chr.16: 30757615-30757615 on Build GRCh38
rs576593404 Chr.16: 30757623-30757623 on Build GRCh38
rs932095063 Chr.16: 30757646-30757646 on Build GRCh38
rs756688945 Chr.16: 30757610-30757610 on Build GRCh38
rs746924858 Chr.16: 30757641-30757641 on Build GRCh38
rs929074119 Chr.16: 30757593-30757593 on Build GRCh38
rs751109759 Chr.16: 30757666-30757666 on Build GRCh38
rs753883532 Chr.16: 30757469-30757469 on Build GRCh38
rs772844089 Chr.16: 30757655-30757655 on Build GRCh38
rs759709742 Chr.16: 30757526-30757526 on Build GRCh38
rs1048703 Chr.16: 30757584-30757584 on Build GRCh38

View Full Product Details