Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 48465376-48465639 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATRIP
Entrez Gene ID
84126
Gene Name
ATR interacting protein
Gene Aliases
-
UniGene
Hs.694840
Gene Chromosome Location
Chr.3: 48446710-48465655 on Build GRCh38
Transcripts
NM_001271022.1 NM_001271023.1 NM_032166.3 NM_130384.2 NM_007248.3 NM_016381.5 NM_033629.4

Gene Symbol
TREX1
Entrez Gene ID
11277
Gene Name
three prime repair exonuclease 1
Gene Aliases
AGS1, CRV, DRN3, HERNS
UniGene
Hs.707026
Gene Chromosome Location
Chr.3: 48465520-48467645 on Build GRCh38
Transcripts
NM_001271022.1 NM_001271023.1 NM_032166.3 NM_130384.2 NM_007248.3 NM_016381.5 NM_033629.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1753250 c.2375G>A p.*792* Chr.3: 48465553-48465553 on Build GRCh38
6342704 c.2330G>T p.C777F Chr.3: 48465508-48465508 on Build GRCh38
3070379 c.2333C>T p.A778V Chr.3: 48465511-48465511 on Build GRCh38
730662 c.2348T>C p.V783A Chr.3: 48465526-48465526 on Build GRCh38
1309215 c.2356C>G p.P786A Chr.3: 48465534-48465534 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs939689929 Chr.3: 48465422-48465422 on Build GRCh38
rs377554132 Chr.3: 48465507-48465507 on Build GRCh38
rs773479091 Chr.3: 48465524-48465524 on Build GRCh38
rs527457931 Chr.3: 48465434-48465434 on Build GRCh38
rs776600109 Chr.3: 48465521-48465521 on Build GRCh38
rs977794304 Chr.3: 48465408-48465408 on Build GRCh38
rs752225103 Chr.3: 48465566-48465566 on Build GRCh38
rs1012118479 Chr.3: 48465599-48465599 on Build GRCh38
rs995326315 Chr.3: 48465441-48465441 on Build GRCh38
rs374648324 Chr.3: 48465503-48465503 on Build GRCh38
rs751472086 Chr.3: 48465558-48465558 on Build GRCh38
rs921504605 Chr.3: 48465402-48465402 on Build GRCh38
rs374400972 Chr.3: 48465537-48465537 on Build GRCh38
rs556531421 Chr.3: 48465411-48465411 on Build GRCh38
rs759414382 Chr.3: 48465559-48465559 on Build GRCh38
rs772624567 Chr.3: 48465513-48465513 on Build GRCh38
rs201794728 Chr.3: 48465515-48465515 on Build GRCh38
rs548746400 Chr.3: 48465457-48465457 on Build GRCh38
rs755752189 Chr.3: 48465600-48465600 on Build GRCh38
rs112793296 Chr.3: 48465413-48465413 on Build GRCh38
rs921473259 Chr.3: 48465598-48465598 on Build GRCh38
rs370310292 Chr.3: 48465468-48465468 on Build GRCh38
rs752344920 Chr.3: 48465482-48465482 on Build GRCh38
rs1022194931 Chr.3: 48465601-48465601 on Build GRCh38
rs538134713 Chr.3: 48465517-48465517 on Build GRCh38
rs761933313 Chr.3: 48465522-48465522 on Build GRCh38
rs750667408 Chr.3: 48465598-48465601 on Build GRCh38
rs765487326 Chr.3: 48465474-48465475 on Build GRCh38
rs898272237 Chr.3: 48465432-48465432 on Build GRCh38
rs967843729 Chr.3: 48465610-48465610 on Build GRCh38
rs766287947 Chr.3: 48465458-48465458 on Build GRCh38
rs755805292 Chr.3: 48465497-48465497 on Build GRCh38
rs967896248 Chr.3: 48465407-48465407 on Build GRCh38
rs751462232 Chr.3: 48465463-48465463 on Build GRCh38
rs532795461 Chr.3: 48465406-48465406 on Build GRCh38
rs547922447 Chr.3: 48465427-48465445 on Build GRCh38
rs780554074 Chr.3: 48465514-48465514 on Build GRCh38
rs962768303 Chr.3: 48465580-48465580 on Build GRCh38
rs767538488 Chr.3: 48465565-48465565 on Build GRCh38
rs780596477 Chr.3: 48465470-48465470 on Build GRCh38
rs762674869 Chr.3: 48465536-48465536 on Build GRCh38
rs750184297 Chr.3: 48465445-48465445 on Build GRCh38
rs141960118 Chr.3: 48465512-48465512 on Build GRCh38

View Full Product Details