Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 90716701-90716956 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BARHL2
Entrez Gene ID
343472
Gene Name
BarH like homeobox 2
Gene Aliases
-
UniGene
Hs.451956
Gene Chromosome Location
Chr.1: 90712022-90717237 on Build GRCh38
Transcripts
NM_020063.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6609947 c.296C>T p.A99V Chr.1: 90716900-90716900 on Build GRCh38
6037167 c.281C>A p.P94Q Chr.1: 90716915-90716915 on Build GRCh38
4627617 c.474A>G p.A158A Chr.1: 90716722-90716722 on Build GRCh38
6044702 c.375_376insC p.Q126fs*20 Chr.1: 90716820-90716821 on Build GRCh38
1293275 c.485_486CC>TT p.T162I Chr.1: 90716710-90716711 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs534277570 Chr.1: 90716839-90716839 on Build GRCh38
rs763642668 Chr.1: 90716905-90716907 on Build GRCh38
rs752327071 Chr.1: 90716825-90716825 on Build GRCh38
rs767961641 Chr.1: 90716793-90716793 on Build GRCh38
rs375986324 Chr.1: 90716894-90716894 on Build GRCh38
rs771807021 Chr.1: 90716741-90716741 on Build GRCh38
rs775419471 Chr.1: 90716758-90716758 on Build GRCh38
rs763565738 Chr.1: 90716838-90716858 on Build GRCh38
rs758033710 Chr.1: 90716911-90716911 on Build GRCh38
rs546122444 Chr.1: 90716830-90716830 on Build GRCh38
rs761069597 Chr.1: 90716802-90716802 on Build GRCh38
rs768831855 Chr.1: 90716848-90716848 on Build GRCh38
rs770646227 Chr.1: 90716838-90716838 on Build GRCh38
rs760087699 Chr.1: 90716897-90716897 on Build GRCh38
rs1026844512 Chr.1: 90716728-90716728 on Build GRCh38
rs751178049 Chr.1: 90716926-90716926 on Build GRCh38
rs576963559 Chr.1: 90716826-90716826 on Build GRCh38
rs902811193 Chr.1: 90716864-90716864 on Build GRCh38
rs555932723 Chr.1: 90716840-90716840 on Build GRCh38
rs778419577 Chr.1: 90716822-90716822 on Build GRCh38
rs748303673 Chr.1: 90716825-90716845 on Build GRCh38
rs757038922 Chr.1: 90716930-90716930 on Build GRCh38
rs774612049 Chr.1: 90716856-90716856 on Build GRCh38
rs777677418 Chr.1: 90716747-90716747 on Build GRCh38
rs752275712 Chr.1: 90716906-90716906 on Build GRCh38
rs773314962 Chr.1: 90716880-90716880 on Build GRCh38
rs532560427 Chr.1: 90716824-90716825 on Build GRCh38
rs746893395 Chr.1: 90716749-90716749 on Build GRCh38
rs756187036 Chr.1: 90716936-90716936 on Build GRCh38
rs1020354585 Chr.1: 90716805-90716805 on Build GRCh38
rs763033411 Chr.1: 90716776-90716776 on Build GRCh38
rs774665217 Chr.1: 90716788-90716788 on Build GRCh38
rs772479464 Chr.1: 90716875-90716875 on Build GRCh38
rs750866481 Chr.1: 90716798-90716798 on Build GRCh38
rs764698944 Chr.1: 90716900-90716900 on Build GRCh38
rs770262529 Chr.1: 90716828-90716845 on Build GRCh38
rs138592538 Chr.1: 90716755-90716755 on Build GRCh38
rs574132317 Chr.1: 90716887-90716887 on Build GRCh38
rs538406922 Chr.1: 90716915-90716915 on Build GRCh38
rs746880153 Chr.1: 90716834-90716834 on Build GRCh38
rs538941381 Chr.1: 90716739-90716739 on Build GRCh38
rs768862477 Chr.1: 90716777-90716777 on Build GRCh38
rs1041397679 Chr.1: 90716858-90716858 on Build GRCh38
rs762120879 Chr.1: 90716874-90716874 on Build GRCh38
rs551290584 Chr.1: 90716731-90716731 on Build GRCh38
rs771363898 Chr.1: 90716845-90716846 on Build GRCh38
rs564798803 Chr.1: 90716915-90716916 on Build GRCh38
rs766681566 Chr.1: 90716895-90716895 on Build GRCh38
rs780896846 Chr.1: 90716933-90716933 on Build GRCh38
rs754303290 Chr.1: 90716813-90716813 on Build GRCh38
rs1049082296 Chr.1: 90716824-90716824 on Build GRCh38
rs932161610 Chr.1: 90716827-90716827 on Build GRCh38
rs745768649 Chr.1: 90716934-90716934 on Build GRCh38
rs768652442 Chr.1: 90716825-90716827 on Build GRCh38
rs774963586 Chr.1: 90716904-90716905 on Build GRCh38
rs773419051 Chr.1: 90716833-90716834 on Build GRCh38
rs930755540 Chr.1: 90716907-90716907 on Build GRCh38
rs566918058 Chr.1: 90716821-90716821 on Build GRCh38
rs551753021 Chr.1: 90716810-90716810 on Build GRCh38
rs747119215 Chr.1: 90716819-90716820 on Build GRCh38
rs988506022 Chr.1: 90716786-90716786 on Build GRCh38
rs762173988 Chr.1: 90716792-90716792 on Build GRCh38
rs960204197 Chr.1: 90716779-90716779 on Build GRCh38

View Full Product Details