Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 128259915-128260182 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GOLGA2
Entrez Gene ID
2801
Gene Name
golgin A2
Gene Aliases
GM130
UniGene
Hs.155827
Gene Chromosome Location
Chr.9: 128255829-128276007 on Build GRCh38
Transcripts
NM_004486.4 XM_005251930.1 XM_005251931.1 XM_005251932.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6672502 c.1692G>T p.M564I Chr.9: 128260175-128260175 on Build GRCh38
7311347 c.1723C>T p.H575Y Chr.9: 128260144-128260144 on Build GRCh38
6672501 c.1727T>G p.V576G Chr.9: 128260140-128260140 on Build GRCh38
3904389 c.1771C>T p.L591L Chr.9: 128260096-128260096 on Build GRCh38
381123 c.1755C>G p.G585G Chr.9: 128260112-128260112 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs750765144 Chr.9: 128260100-128260100 on Build GRCh38
rs768763105 Chr.9: 128260152-128260152 on Build GRCh38
rs199538572 Chr.9: 128260111-128260111 on Build GRCh38
rs376863046 Chr.9: 128260041-128260041 on Build GRCh38
rs200653956 Chr.9: 128260068-128260068 on Build GRCh38
rs369587258 Chr.9: 128260042-128260042 on Build GRCh38
rs541697888 Chr.9: 128260037-128260037 on Build GRCh38
rs776765819 Chr.9: 128260084-128260085 on Build GRCh38
rs940898192 Chr.9: 128259958-128259958 on Build GRCh38
rs770450224 Chr.9: 128260133-128260133 on Build GRCh38
rs759522002 Chr.9: 128260100-128260102 on Build GRCh38
rs767951870 Chr.9: 128260091-128260091 on Build GRCh38
rs746641261 Chr.9: 128260131-128260131 on Build GRCh38
rs142411612 Chr.9: 128260162-128260162 on Build GRCh38
rs757615879 Chr.9: 128260090-128260090 on Build GRCh38
rs775404277 Chr.9: 128260143-128260143 on Build GRCh38
rs780988029 Chr.9: 128260136-128260136 on Build GRCh38
rs759502737 Chr.9: 128260067-128260067 on Build GRCh38
rs201243209 Chr.9: 128260108-128260108 on Build GRCh38
rs751797429 Chr.9: 128260077-128260077 on Build GRCh38
rs748473256 Chr.9: 128260039-128260039 on Build GRCh38
rs1034965776 Chr.9: 128260076-128260076 on Build GRCh38
rs148778498 Chr.9: 128260120-128260120 on Build GRCh38
rs145670991 Chr.9: 128260023-128260023 on Build GRCh38
rs776471946 Chr.9: 128260045-128260045 on Build GRCh38
rs764293854 Chr.9: 128260072-128260072 on Build GRCh38
rs182615718 Chr.9: 128260033-128260033 on Build GRCh38
rs751878112 Chr.9: 128260032-128260032 on Build GRCh38
rs1003804193 Chr.9: 128259959-128259959 on Build GRCh38
rs374197968 Chr.9: 128260139-128260139 on Build GRCh38
rs1015590799 Chr.9: 128260008-128260008 on Build GRCh38
rs757910820 Chr.9: 128260112-128260112 on Build GRCh38
rs956818908 Chr.9: 128260078-128260078 on Build GRCh38
rs373870855 Chr.9: 128260151-128260151 on Build GRCh38
rs765799898 Chr.9: 128260085-128260105 on Build GRCh38
rs775563471 Chr.9: 128260070-128260070 on Build GRCh38
rs778584663 Chr.9: 128260040-128260040 on Build GRCh38
rs745605906 Chr.9: 128260137-128260137 on Build GRCh38
rs975124711 Chr.9: 128260122-128260122 on Build GRCh38
rs770736942 Chr.9: 128260044-128260044 on Build GRCh38
rs756699959 Chr.9: 128260104-128260104 on Build GRCh38

View Full Product Details