Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 32534498-32534739 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DYNC1LI1
Entrez Gene ID
51143
Gene Name
dynein cytoplasmic 1 light intermediate chain 1
Gene Aliases
DLC-A, DNCLI1, LIC1
UniGene
Hs.529495
Gene Chromosome Location
Chr.3: 32525971-32570874 on Build GRCh38
Transcripts
NM_016141.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5864309 c.904C>T p.Q302* Chr.3: 32534575-32534575 on Build GRCh38
280669 c.865G>T p.E289* Chr.3: 32534614-32534614 on Build GRCh38
6650614 c.962T>C p.V321A Chr.3: 32534517-32534517 on Build GRCh38
1043655 c.900C>T p.I300I Chr.3: 32534579-32534579 on Build GRCh38
3823667 c.857C>T p.S286L Chr.3: 32534622-32534622 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs62252795 Chr.3: 32534691-32534691 on Build GRCh38
rs201749401 Chr.3: 32534619-32534619 on Build GRCh38
rs1038013721 Chr.3: 32534635-32534635 on Build GRCh38
rs749564113 Chr.3: 32534598-32534598 on Build GRCh38
rs750122388 Chr.3: 32534553-32534553 on Build GRCh38
rs760929356 Chr.3: 32534567-32534567 on Build GRCh38
rs955460173 Chr.3: 32534675-32534675 on Build GRCh38
rs753917450 Chr.3: 32534657-32534657 on Build GRCh38
rs780877304 Chr.3: 32534576-32534576 on Build GRCh38
rs753496869 Chr.3: 32534666-32534666 on Build GRCh38
rs773083907 Chr.3: 32534670-32534670 on Build GRCh38
rs766347359 Chr.3: 32534682-32534682 on Build GRCh38
rs768723065 Chr.3: 32534656-32534656 on Build GRCh38
rs150042247 Chr.3: 32534618-32534618 on Build GRCh38
rs779530074 Chr.3: 32534627-32534627 on Build GRCh38
rs766182253 Chr.3: 32534566-32534566 on Build GRCh38
rs775655183 Chr.3: 32534646-32534646 on Build GRCh38
rs778230309 Chr.3: 32534581-32534581 on Build GRCh38
rs746012118 Chr.3: 32534631-32534631 on Build GRCh38
rs772465213 Chr.3: 32534535-32534535 on Build GRCh38
rs775851300 Chr.3: 32534538-32534538 on Build GRCh38
rs569367799 Chr.3: 32534578-32534578 on Build GRCh38
rs762982173 Chr.3: 32534557-32534557 on Build GRCh38
rs766534167 Chr.3: 32534639-32534639 on Build GRCh38
rs372315791 Chr.3: 32534654-32534654 on Build GRCh38
rs151322643 Chr.3: 32534568-32534568 on Build GRCh38
rs183663455 Chr.3: 32534693-32534693 on Build GRCh38
rs1002674557 Chr.3: 32534707-32534707 on Build GRCh38
rs192375768 Chr.3: 32534683-32534683 on Build GRCh38
rs761760806 Chr.3: 32534541-32534541 on Build GRCh38
rs755651756 Chr.3: 32534579-32534579 on Build GRCh38
rs184025487 Chr.3: 32534545-32534545 on Build GRCh38
rs907041591 Chr.3: 32534706-32534706 on Build GRCh38
rs200552230 Chr.3: 32534674-32534674 on Build GRCh38

View Full Product Details