Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 78833507-78833766 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BMP2K
Entrez Gene ID
55589
Gene Name
BMP2 inducible kinase
Gene Aliases
BIKE, HRIHFB2017
UniGene
Hs.146551
Gene Chromosome Location
Chr.4: 78776378-78916365 on Build GRCh38
Transcripts
NM_017593.3 NM_198892.1 XM_005263117.1 XM_011532102.1 XM_017008381.1 XM_017008382.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6611920 c.403+7_403+8insT p.? Chr.4: 78833694-78833695 on Build GRCh38
6611930 c.348_349insA p.N117fs*4 Chr.4: 78833632-78833633 on Build GRCh38
4126167 c.400C>T p.R134* Chr.4: 78833684-78833684 on Build GRCh38
4126169 c.400C>T p.R134* Chr.4: 78833684-78833684 on Build GRCh38
318926 c.313C>T p.H105Y Chr.4: 78833597-78833597 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs776105161 Chr.4: 78833711-78833712 on Build GRCh38
rs768793996 Chr.4: 78833574-78833574 on Build GRCh38
rs748375720 Chr.4: 78833584-78833584 on Build GRCh38
rs1051923338 Chr.4: 78833715-78833715 on Build GRCh38
rs373570708 Chr.4: 78833544-78833544 on Build GRCh38
rs774904128 Chr.4: 78833625-78833625 on Build GRCh38
rs769669826 Chr.4: 78833716-78833716 on Build GRCh38
rs200365695 Chr.4: 78833604-78833604 on Build GRCh38
rs761810376 Chr.4: 78833676-78833676 on Build GRCh38
rs1033388903 Chr.4: 78833547-78833547 on Build GRCh38
rs150783429 Chr.4: 78833588-78833588 on Build GRCh38
rs758640857 Chr.4: 78833694-78833694 on Build GRCh38
rs768165615 Chr.4: 78833574-78833574 on Build GRCh38
rs796077213 Chr.4: 78833563-78833564 on Build GRCh38
rs774933681 Chr.4: 78833720-78833720 on Build GRCh38
rs759453115 Chr.4: 78833616-78833616 on Build GRCh38
rs749258535 Chr.4: 78833567-78833567 on Build GRCh38
rs149913140 Chr.4: 78833668-78833668 on Build GRCh38
rs373652417 Chr.4: 78833712-78833712 on Build GRCh38
rs756564717 Chr.4: 78833542-78833542 on Build GRCh38
rs775438894 Chr.4: 78833562-78833563 on Build GRCh38
rs776647701 Chr.4: 78833614-78833614 on Build GRCh38
rs769587908 Chr.4: 78833685-78833685 on Build GRCh38
rs112619803 Chr.4: 78833563-78833563 on Build GRCh38
rs762574402 Chr.4: 78833629-78833629 on Build GRCh38
rs114119862 Chr.4: 78833578-78833578 on Build GRCh38
rs370858578 Chr.4: 78833711-78833711 on Build GRCh38
rs750747429 Chr.4: 78833653-78833653 on Build GRCh38
rs778169576 Chr.4: 78833708-78833708 on Build GRCh38
rs772459543 Chr.4: 78833585-78833585 on Build GRCh38
rs748285805 Chr.4: 78833687-78833687 on Build GRCh38
rs201025295 Chr.4: 78833639-78833639 on Build GRCh38
rs368136183 Chr.4: 78833617-78833617 on Build GRCh38
rs1002211420 Chr.4: 78833675-78833675 on Build GRCh38
rs756474729 Chr.4: 78833659-78833659 on Build GRCh38
rs139141439 Chr.4: 78833660-78833660 on Build GRCh38
rs773117319 Chr.4: 78833587-78833587 on Build GRCh38
rs754354277 Chr.4: 78833667-78833667 on Build GRCh38
rs568742609 Chr.4: 78833563-78833564 on Build GRCh38
rs745790512 Chr.4: 78833713-78833713 on Build GRCh38
rs773108508 Chr.4: 78833735-78833735 on Build GRCh38
rs746818379 Chr.4: 78833596-78833596 on Build GRCh38

View Full Product Details