Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 99450637-99450874 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATP5J2-PTCD1
Entrez Gene ID
100526740
Gene Name
ATP5J2-PTCD1 readthrough
Gene Aliases
-
UniGene
Hs.632313
Gene Chromosome Location
Chr.7: 99416739-99466201 on Build GRCh38
Transcripts
NM_001198879.1 NM_001081559.2 NM_001318160.1 NM_001318161.1 NM_001318162.1 NM_006693.3 XM_011515755.2 XM_011515756.2 XM_011515757.2 XM_017011700.1 XM_017011701.1 XM_017011702.1 XM_017011703.1

Gene Symbol
CPSF4
Entrez Gene ID
10898
Gene Name
cleavage and polyadenylation specific factor 4
Gene Aliases
CPSF30, NAR, NEB-1, NEB1
UniGene
Hs.489287
Gene Chromosome Location
Chr.7: 99438920-99457377 on Build GRCh38
Transcripts
NM_001198879.1 NM_001081559.2 NM_001318160.1 NM_001318161.1 NM_001318162.1 NM_006693.3 XM_011515755.2 XM_011515756.2 XM_011515757.2 XM_017011700.1 XM_017011701.1 XM_017011702.1 XM_017011703.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1093807 c.457G>A p.V153M Chr.7: 99450755-99450755 on Build GRCh38
6419744 c.460G>A p.G154R Chr.7: 99450758-99450758 on Build GRCh38
7215249 c.423G>A p.R141R Chr.7: 99450721-99450721 on Build GRCh38
7400641 c.434G>C p.R145T Chr.7: 99450732-99450732 on Build GRCh38
1249062 c.454C>T p.L152F Chr.7: 99450752-99450752 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs758998260 Chr.7: 99450814-99450814 on Build GRCh38
rs765727367 Chr.7: 99450674-99450674 on Build GRCh38
rs550355025 Chr.7: 99450855-99450855 on Build GRCh38
rs747222779 Chr.7: 99450834-99450834 on Build GRCh38
rs761619358 Chr.7: 99450823-99450823 on Build GRCh38
rs752817358 Chr.7: 99450694-99450706 on Build GRCh38
rs374028074 Chr.7: 99450815-99450815 on Build GRCh38
rs754489272 Chr.7: 99450689-99450689 on Build GRCh38
rs752654171 Chr.7: 99450736-99450736 on Build GRCh38
rs766025488 Chr.7: 99450807-99450807 on Build GRCh38
rs188284424 Chr.7: 99450766-99450766 on Build GRCh38
rs777553781 Chr.7: 99450754-99450754 on Build GRCh38
rs780176480 Chr.7: 99450768-99450768 on Build GRCh38
rs561665792 Chr.7: 99450818-99450818 on Build GRCh38
rs757048228 Chr.7: 99450832-99450832 on Build GRCh38
rs763295609 Chr.7: 99450806-99450806 on Build GRCh38
rs2280600 Chr.7: 99450809-99450809 on Build GRCh38
rs900388282 Chr.7: 99450687-99450687 on Build GRCh38
rs762216649 Chr.7: 99450673-99450673 on Build GRCh38
rs766003852 Chr.7: 99450683-99450683 on Build GRCh38
rs200509947 Chr.7: 99450767-99450767 on Build GRCh38
rs45609448 Chr.7: 99450680-99450680 on Build GRCh38
rs560365724 Chr.7: 99450780-99450780 on Build GRCh38
rs755040956 Chr.7: 99450839-99450839 on Build GRCh38
rs777322552 Chr.7: 99450667-99450667 on Build GRCh38
rs371928827 Chr.7: 99450755-99450755 on Build GRCh38
rs768876531 Chr.7: 99450775-99450775 on Build GRCh38
rs201191297 Chr.7: 99450677-99450677 on Build GRCh38
rs544307868 Chr.7: 99450684-99450684 on Build GRCh38
rs753633688 Chr.7: 99450827-99450827 on Build GRCh38
rs780269981 Chr.7: 99450833-99450833 on Build GRCh38
rs182798242 Chr.7: 99450743-99450743 on Build GRCh38
rs781085938 Chr.7: 99450698-99450698 on Build GRCh38
rs773611678 Chr.7: 99450803-99450803 on Build GRCh38
rs199773463 Chr.7: 99450820-99450820 on Build GRCh38
rs747085181 Chr.7: 99450769-99450769 on Build GRCh38
rs781434272 Chr.7: 99450841-99450841 on Build GRCh38
rs35303224 Chr.7: 99450781-99450781 on Build GRCh38
rs767513910 Chr.7: 99450836-99450836 on Build GRCh38
rs770224170 Chr.7: 99450790-99450790 on Build GRCh38
rs887880870 Chr.7: 99450774-99450774 on Build GRCh38

View Full Product Details