Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 197040561-197040814 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
F13B
Entrez Gene ID
2165
Gene Name
coagulation factor XIII B chain
Gene Aliases
FXIIIB
UniGene
Hs.435782
Gene Chromosome Location
Chr.1: 197038741-197067267 on Build GRCh38
Transcripts
NM_001994.2 XM_011509283.2 XM_011509284.2 XM_011509286.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4452487 c.1912A>T p.R638* Chr.1: 197040562-197040562 on Build GRCh38
901614 c.1855G>T p.D619Y Chr.1: 197040619-197040619 on Build GRCh38
6657694 c.1755T>A p.S585S Chr.1: 197040719-197040719 on Build GRCh38
6052684 c.1886G>A p.G629E Chr.1: 197040588-197040588 on Build GRCh38
1473206 c.1762G>A p.E588K Chr.1: 197040712-197040712 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs866221600 Chr.1: 197040729-197040729 on Build GRCh38
rs769535126 Chr.1: 197040747-197040747 on Build GRCh38
rs750551312 Chr.1: 197040592-197040592 on Build GRCh38
rs544335603 Chr.1: 197040693-197040693 on Build GRCh38
rs111517419 Chr.1: 197040772-197040772 on Build GRCh38
rs774352966 Chr.1: 197040680-197040680 on Build GRCh38
rs760721695 Chr.1: 197040762-197040767 on Build GRCh38
rs959081263 Chr.1: 197040771-197040771 on Build GRCh38
rs770292850 Chr.1: 197040643-197040643 on Build GRCh38
rs756826341 Chr.1: 197040733-197040733 on Build GRCh38
rs780628616 Chr.1: 197040752-197040752 on Build GRCh38
rs775772881 Chr.1: 197040758-197040761 on Build GRCh38
rs746518541 Chr.1: 197040631-197040631 on Build GRCh38
rs774924051 Chr.1: 197040770-197040770 on Build GRCh38
rs780903820 Chr.1: 197040653-197040653 on Build GRCh38
rs759350789 Chr.1: 197040752-197040752 on Build GRCh38
rs761964408 Chr.1: 197040691-197040691 on Build GRCh38
rs556380704 Chr.1: 197040741-197040743 on Build GRCh38
rs768845338 Chr.1: 197040765-197040765 on Build GRCh38
rs758392032 Chr.1: 197040728-197040728 on Build GRCh38
rs575018842 Chr.1: 197040751-197040752 on Build GRCh38
rs765477781 Chr.1: 197040649-197040649 on Build GRCh38
rs5998 Chr.1: 197040668-197040668 on Build GRCh38
rs770302077 Chr.1: 197040666-197040666 on Build GRCh38
rs749655588 Chr.1: 197040757-197040757 on Build GRCh38
rs760783227 Chr.1: 197040721-197040721 on Build GRCh38
rs780516724 Chr.1: 197040739-197040739 on Build GRCh38
rs745562332 Chr.1: 197040743-197040743 on Build GRCh38
rs777784021 Chr.1: 197040730-197040730 on Build GRCh38
rs770951653 Chr.1: 197040671-197040671 on Build GRCh38
rs758301404 Chr.1: 197040610-197040610 on Build GRCh38
rs908534762 Chr.1: 197040640-197040640 on Build GRCh38
rs766164044 Chr.1: 197040718-197040718 on Build GRCh38
rs368242266 Chr.1: 197040701-197040701 on Build GRCh38
rs375961962 Chr.1: 197040659-197040659 on Build GRCh38
rs778632077 Chr.1: 197040608-197040608 on Build GRCh38
rs751388812 Chr.1: 197040732-197040732 on Build GRCh38
rs777215791 Chr.1: 197040623-197040623 on Build GRCh38
rs748133917 Chr.1: 197040609-197040609 on Build GRCh38
rs147163101 Chr.1: 197040720-197040720 on Build GRCh38
rs760866432 Chr.1: 197040702-197040702 on Build GRCh38
rs762185550 Chr.1: 197040782-197040782 on Build GRCh38
rs201438197 Chr.1: 197040597-197040597 on Build GRCh38
rs763591423 Chr.1: 197040669-197040669 on Build GRCh38
rs769200824 Chr.1: 197040672-197040672 on Build GRCh38

View Full Product Details