Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 68839939-68840172 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EFNB1
Entrez Gene ID
1947
Gene Name
ephrin B1
Gene Aliases
CFND, CFNS, EFB1, EFL3, EPLG2, Elk-L, LERK2
UniGene
Hs.144700
Gene Chromosome Location
Chr.X: 68828997-68842164 on Build GRCh38
Transcripts
NM_004429.4

SNP Detail

SNP ID Chromosome Location
rs766918704 Chr.X: 68840068-68840068 on Build GRCh38
rs7889678 Chr.X: 68839975-68839975 on Build GRCh38
rs763881049 Chr.X: 68840020-68840020 on Build GRCh38
rs1050128606 Chr.X: 68840150-68840150 on Build GRCh38
rs779526112 Chr.X: 68839971-68839971 on Build GRCh38
rs143162125 Chr.X: 68840000-68840000 on Build GRCh38
rs780539184 Chr.X: 68840127-68840127 on Build GRCh38
rs200409169 Chr.X: 68840057-68840057 on Build GRCh38
rs974195266 Chr.X: 68840044-68840044 on Build GRCh38
rs985853678 Chr.X: 68840128-68840128 on Build GRCh38
rs371670091 Chr.X: 68840119-68840119 on Build GRCh38
rs762345929 Chr.X: 68839986-68839986 on Build GRCh38
rs149648624 Chr.X: 68840059-68840059 on Build GRCh38
rs1057519035 Chr.X: 68840021-68840021 on Build GRCh38
rs377269776 Chr.X: 68839976-68839976 on Build GRCh38
rs546056194 Chr.X: 68840055-68840055 on Build GRCh38
rs756265628 Chr.X: 68840076-68840076 on Build GRCh38
rs762694943 Chr.X: 68840009-68840009 on Build GRCh38
rs16989105 Chr.X: 68840026-68840026 on Build GRCh38
rs959897469 Chr.X: 68839966-68839966 on Build GRCh38
rs754048678 Chr.X: 68840079-68840079 on Build GRCh38
rs748653660 Chr.X: 68840122-68840122 on Build GRCh38
rs978471287 Chr.X: 68840030-68840030 on Build GRCh38
rs754041672 Chr.X: 68840095-68840095 on Build GRCh38
rs759040680 Chr.X: 68840125-68840125 on Build GRCh38
rs746729523 Chr.X: 68840093-68840094 on Build GRCh38
rs767884649 Chr.X: 68840004-68840004 on Build GRCh38
rs755331344 Chr.X: 68840101-68840101 on Build GRCh38
rs150883256 Chr.X: 68840025-68840025 on Build GRCh38
rs147599832 Chr.X: 68839969-68839969 on Build GRCh38

View Full Product Details