Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 38815632-38815873 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ECH1
Entrez Gene ID
1891
Gene Name
enoyl-CoA hydratase 1
Gene Aliases
HPXEL
UniGene
Hs.196176
Gene Chromosome Location
Chr.19: 38815422-38831857 on Build GRCh38
Transcripts
NM_001398.2 XM_017026448.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6651333 c.883-2A>G p.? Chr.19: 38815719-38815719 on Build GRCh38
3223004 c.966C>T p.T322T Chr.19: 38815634-38815634 on Build GRCh38
7090122 c.961A>G p.K321E Chr.19: 38815639-38815639 on Build GRCh38
5427076 c.883-6T>C p.? Chr.19: 38815723-38815723 on Build GRCh38
3533574 c.887C>T p.S296F Chr.19: 38815713-38815713 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs759998789 Chr.19: 38815741-38815741 on Build GRCh38
rs745668987 Chr.19: 38815703-38815703 on Build GRCh38
rs552130567 Chr.19: 38815845-38815845 on Build GRCh38
rs933038881 Chr.19: 38815837-38815837 on Build GRCh38
rs770617489 Chr.19: 38815679-38815679 on Build GRCh38
rs374092073 Chr.19: 38815726-38815726 on Build GRCh38
rs1038846541 Chr.19: 38815733-38815733 on Build GRCh38
rs777560195 Chr.19: 38815839-38815840 on Build GRCh38
rs192745108 Chr.19: 38815835-38815835 on Build GRCh38
rs773981477 Chr.19: 38815831-38815831 on Build GRCh38
rs552518184 Chr.19: 38815807-38815807 on Build GRCh38
rs769380241 Chr.19: 38815710-38815710 on Build GRCh38
rs370357377 Chr.19: 38815840-38815840 on Build GRCh38
rs777540153 Chr.19: 38815666-38815666 on Build GRCh38
rs144521764 Chr.19: 38815670-38815670 on Build GRCh38
rs748993959 Chr.19: 38815820-38815820 on Build GRCh38
rs755934996 Chr.19: 38815814-38815814 on Build GRCh38
rs779650677 Chr.19: 38815817-38815817 on Build GRCh38
rs764291702 Chr.19: 38815748-38815748 on Build GRCh38
rs567275196 Chr.19: 38815713-38815713 on Build GRCh38
rs377138593 Chr.19: 38815737-38815737 on Build GRCh38
rs2075588 Chr.19: 38815785-38815785 on Build GRCh38
rs765745607 Chr.19: 38815847-38815847 on Build GRCh38
rs374422461 Chr.19: 38815841-38815841 on Build GRCh38
rs569568697 Chr.19: 38815745-38815745 on Build GRCh38
rs765200783 Chr.19: 38815660-38815660 on Build GRCh38
rs775841218 Chr.19: 38815850-38815850 on Build GRCh38
rs758250409 Chr.19: 38815672-38815674 on Build GRCh38
rs530927065 Chr.19: 38815791-38815791 on Build GRCh38
rs530366183 Chr.19: 38815783-38815783 on Build GRCh38
rs781007249 Chr.19: 38815766-38815766 on Build GRCh38
rs974345677 Chr.19: 38815797-38815797 on Build GRCh38
rs768040303 Chr.19: 38815822-38815822 on Build GRCh38
rs369819365 Chr.19: 38815738-38815738 on Build GRCh38
rs761304441 Chr.19: 38815731-38815731 on Build GRCh38
rs758384942 Chr.19: 38815747-38815747 on Build GRCh38
rs751445868 Chr.19: 38815751-38815751 on Build GRCh38
rs987603672 Chr.19: 38815699-38815699 on Build GRCh38
rs868174591 Chr.19: 38815716-38815716 on Build GRCh38
rs374473358 Chr.19: 38815744-38815744 on Build GRCh38
rs913591129 Chr.19: 38815678-38815678 on Build GRCh38
rs1033886239 Chr.19: 38815846-38815846 on Build GRCh38
rs532825433 Chr.19: 38815684-38815684 on Build GRCh38
rs11549272 Chr.19: 38815697-38815697 on Build GRCh38
rs773574397 Chr.19: 38815729-38815729 on Build GRCh38
rs763300655 Chr.19: 38815851-38815851 on Build GRCh38
rs754195436 Chr.19: 38815740-38815740 on Build GRCh38
rs887132626 Chr.19: 38815784-38815784 on Build GRCh38
rs369437189 Chr.19: 38815658-38815658 on Build GRCh38
rs559979174 Chr.19: 38815773-38815773 on Build GRCh38
rs762410107 Chr.19: 38815717-38815717 on Build GRCh38

View Full Product Details