Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 2431594-2431863 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LMNB2
Entrez Gene ID
84823
Gene Name
lamin B2
Gene Aliases
EPM9, LAMB2, LMN2
UniGene
Hs.538286
Gene Chromosome Location
Chr.19: 2428165-2456968 on Build GRCh38
Transcripts
NM_032737.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2749485 c.1574C>T p.T525M Chr.19: 2431859-2431859 on Build GRCh38
4076476 c.1707G>A p.G569G Chr.19: 2431602-2431602 on Build GRCh38
994273 c.1648G>A p.E550K Chr.19: 2431785-2431785 on Build GRCh38
216444 c.1571C>T p.S524L Chr.19: 2431862-2431862 on Build GRCh38
6701836 c.1707C>T p.G569G Chr.19: 2431786-2431786 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs995586631 Chr.19: 2431623-2431624 on Build GRCh38
rs907841277 Chr.19: 2431721-2431721 on Build GRCh38
rs377036714 Chr.19: 2431618-2431618 on Build GRCh38
rs777523411 Chr.19: 2431677-2431677 on Build GRCh38
rs898643557 Chr.19: 2431718-2431718 on Build GRCh38
rs770512914 Chr.19: 2431765-2431766 on Build GRCh38
rs1037741313 Chr.19: 2431743-2431743 on Build GRCh38
rs755478316 Chr.19: 2431768-2431768 on Build GRCh38
rs753113068 Chr.19: 2431817-2431817 on Build GRCh38
rs940259022 Chr.19: 2431714-2431714 on Build GRCh38
rs142082911 Chr.19: 2431626-2431626 on Build GRCh38
rs776163864 Chr.19: 2431648-2431648 on Build GRCh38
rs759051519 Chr.19: 2431706-2431706 on Build GRCh38
rs777128431 Chr.19: 2431696-2431696 on Build GRCh38
rs771771900 Chr.19: 2431792-2431792 on Build GRCh38
rs377753934 Chr.19: 2431800-2431800 on Build GRCh38
rs751633226 Chr.19: 2431812-2431812 on Build GRCh38
rs150787379 Chr.19: 2431632-2431632 on Build GRCh38
rs764021963 Chr.19: 2431753-2431753 on Build GRCh38
rs201873197 Chr.19: 2431619-2431619 on Build GRCh38
rs200972975 Chr.19: 2431689-2431689 on Build GRCh38
rs916303495 Chr.19: 2431687-2431687 on Build GRCh38
rs766983535 Chr.19: 2431735-2431735 on Build GRCh38
rs200332250 Chr.19: 2431744-2431744 on Build GRCh38
rs147619532 Chr.19: 2431795-2431795 on Build GRCh38
rs200122476 Chr.19: 2431665-2431665 on Build GRCh38
rs755132760 Chr.19: 2431813-2431813 on Build GRCh38
rs540663320 Chr.19: 2431715-2431715 on Build GRCh38
rs770562857 Chr.19: 2431701-2431701 on Build GRCh38
rs767398209 Chr.19: 2431671-2431671 on Build GRCh38
rs149924406 Chr.19: 2431785-2431785 on Build GRCh38
rs966453657 Chr.19: 2431636-2431636 on Build GRCh38
rs889836550 Chr.19: 2431729-2431729 on Build GRCh38
rs373868527 Chr.19: 2431690-2431690 on Build GRCh38
rs139202712 Chr.19: 2431653-2431653 on Build GRCh38
rs372581793 Chr.19: 2431786-2431786 on Build GRCh38
rs763677892 Chr.19: 2431722-2431722 on Build GRCh38
rs199507814 Chr.19: 2431755-2431755 on Build GRCh38
rs758739059 Chr.19: 2431762-2431762 on Build GRCh38
rs750289586 Chr.19: 2431809-2431809 on Build GRCh38
rs753609214 Chr.19: 2431754-2431754 on Build GRCh38
rs765459080 Chr.19: 2431759-2431759 on Build GRCh38
rs951618222 Chr.19: 2431734-2431734 on Build GRCh38
rs369838129 Chr.19: 2431816-2431816 on Build GRCh38
rs368075592 Chr.19: 2431676-2431676 on Build GRCh38
rs777932700 Chr.19: 2431622-2431622 on Build GRCh38
rs753332163 Chr.19: 2431763-2431763 on Build GRCh38
rs753636600 Chr.19: 2431679-2431679 on Build GRCh38
rs762650809 Chr.19: 2431662-2431662 on Build GRCh38
rs374358274 Chr.19: 2431826-2431826 on Build GRCh38
rs745499299 Chr.19: 2431787-2431787 on Build GRCh38
rs984094615 Chr.19: 2431748-2431748 on Build GRCh38
rs752662394 Chr.19: 2431672-2431672 on Build GRCh38
rs144445359 Chr.19: 2431776-2431776 on Build GRCh38
rs566466161 Chr.19: 2431838-2431838 on Build GRCh38
rs755926743 Chr.19: 2431621-2431621 on Build GRCh38
rs202017219 Chr.19: 2431680-2431680 on Build GRCh38
rs367739258 Chr.19: 2431694-2431694 on Build GRCh38
rs748469245 Chr.19: 2431698-2431698 on Build GRCh38
rs978070104 Chr.19: 2431670-2431670 on Build GRCh38
rs780317771 Chr.19: 2431692-2431692 on Build GRCh38
rs371265703 Chr.19: 2431695-2431695 on Build GRCh38
rs773901398 Chr.19: 2431703-2431703 on Build GRCh38
rs769107743 Chr.19: 2431652-2431652 on Build GRCh38
rs781566425 Chr.19: 2431775-2431775 on Build GRCh38
rs371087181 Chr.19: 2431664-2431664 on Build GRCh38
rs756697137 Chr.19: 2431821-2431821 on Build GRCh38
rs139812127 Chr.19: 2431620-2431620 on Build GRCh38
rs966777357 Chr.19: 2431760-2431760 on Build GRCh38
rs199916885 Chr.19: 2431716-2431716 on Build GRCh38
rs145444042 Chr.19: 2431811-2431811 on Build GRCh38
rs148138642 Chr.19: 2431715-2431716 on Build GRCh38
rs978396284 Chr.19: 2431769-2431769 on Build GRCh38
rs760189690 Chr.19: 2431794-2431794 on Build GRCh38
rs774978600 Chr.19: 2431793-2431793 on Build GRCh38
rs777181580 Chr.19: 2431661-2431661 on Build GRCh38
rs201618968 Chr.19: 2431806-2431806 on Build GRCh38
rs747149678 Chr.19: 2431765-2431765 on Build GRCh38
rs147230168 Chr.19: 2431627-2431627 on Build GRCh38
rs761076527 Chr.19: 2431651-2431651 on Build GRCh38
rs376849103 Chr.19: 2431741-2431741 on Build GRCh38
rs746627716 Chr.19: 2431759-2431760 on Build GRCh38
rs140533741 Chr.19: 2431807-2431807 on Build GRCh38
rs758984909 Chr.19: 2431669-2431669 on Build GRCh38
rs749496136 Chr.19: 2431625-2431625 on Build GRCh38

View Full Product Details