Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 77464179-77464440 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HMG20A
Entrez Gene ID
10363
Gene Name
high mobility group 20A
Gene Aliases
HMGX1, HMGXB1
UniGene
Hs.69594
Gene Chromosome Location
Chr.15: 77420651-77519899 on Build GRCh38
Transcripts
NM_001304504.1 NM_001304505.1 NM_018200.3 XM_011521158.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1749329 c.229G>A p.E77K Chr.15: 77464379-77464379 on Build GRCh38
3887533 c.165C>T p.L55L Chr.15: 77464315-77464315 on Build GRCh38
3361536 c.126C>T p.A42A Chr.15: 77464276-77464276 on Build GRCh38
4988837 c.127A>G p.T43A Chr.15: 77464277-77464277 on Build GRCh38
97959 c.125C>T p.A42V Chr.15: 77464275-77464275 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs375242827 Chr.15: 77464265-77464265 on Build GRCh38
rs754518003 Chr.15: 77464282-77464282 on Build GRCh38
rs775805602 Chr.15: 77464365-77464365 on Build GRCh38
rs927621947 Chr.15: 77464250-77464250 on Build GRCh38
rs776891578 Chr.15: 77464227-77464227 on Build GRCh38
rs538992863 Chr.15: 77464326-77464326 on Build GRCh38
rs930939773 Chr.15: 77464274-77464274 on Build GRCh38
rs550852751 Chr.15: 77464369-77464369 on Build GRCh38
rs768901087 Chr.15: 77464375-77464375 on Build GRCh38
rs147484814 Chr.15: 77464249-77464249 on Build GRCh38
rs374920143 Chr.15: 77464218-77464218 on Build GRCh38
rs372202469 Chr.15: 77464219-77464219 on Build GRCh38
rs946556270 Chr.15: 77464400-77464400 on Build GRCh38
rs777656083 Chr.15: 77464310-77464310 on Build GRCh38
rs757034031 Chr.15: 77464335-77464335 on Build GRCh38
rs769914968 Chr.15: 77464236-77464236 on Build GRCh38
rs908110754 Chr.15: 77464279-77464279 on Build GRCh38
rs745682274 Chr.15: 77464363-77464363 on Build GRCh38
rs755784117 Chr.15: 77464338-77464339 on Build GRCh38
rs112061352 Chr.15: 77464266-77464266 on Build GRCh38
rs753522168 Chr.15: 77464275-77464275 on Build GRCh38
rs762472420 Chr.15: 77464386-77464386 on Build GRCh38
rs774982088 Chr.15: 77464383-77464383 on Build GRCh38
rs757935496 Chr.15: 77464307-77464307 on Build GRCh38
rs767825320 Chr.15: 77464269-77464269 on Build GRCh38
rs764954436 Chr.15: 77464291-77464291 on Build GRCh38
rs905987899 Chr.15: 77464327-77464327 on Build GRCh38
rs752152965 Chr.15: 77464294-77464294 on Build GRCh38
rs938996794 Chr.15: 77464277-77464277 on Build GRCh38
rs1002020846 Chr.15: 77464342-77464342 on Build GRCh38
rs777707523 Chr.15: 77464215-77464215 on Build GRCh38
rs763142110 Chr.15: 77464259-77464259 on Build GRCh38
rs746135443 Chr.15: 77464231-77464231 on Build GRCh38
rs764648904 Chr.15: 77464262-77464262 on Build GRCh38
rs780917589 Chr.15: 77464348-77464348 on Build GRCh38
rs913815775 Chr.15: 77464384-77464384 on Build GRCh38

View Full Product Details