Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 45805015-45805240 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DNTTIP1
Entrez Gene ID
116092
Gene Name
deoxynucleotidyltransferase, terminal, interacting protein 1
Gene Aliases
C20orf167, Tdif1, dJ447F3.4
UniGene
Hs.472852
Gene Chromosome Location
Chr.20: 45791937-45811427 on Build GRCh38
Transcripts
NM_052951.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4502585 c.616C>T p.R206C Chr.20: 45805158-45805158 on Build GRCh38
4098862 c.604T>C p.W202R Chr.20: 45805146-45805146 on Build GRCh38
6444365 c.625G>A p.E209K Chr.20: 45805167-45805167 on Build GRCh38
5392201 c.652C>T p.R218* Chr.20: 45805194-45805194 on Build GRCh38
6647902 c.636C>A p.T212T Chr.20: 45805178-45805178 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs774068634 Chr.20: 45805122-45805122 on Build GRCh38
rs753642720 Chr.20: 45805161-45805161 on Build GRCh38
rs183196855 Chr.20: 45805110-45805110 on Build GRCh38
rs778512315 Chr.20: 45805184-45805184 on Build GRCh38
rs376375662 Chr.20: 45805123-45805123 on Build GRCh38
rs368814672 Chr.20: 45805118-45805118 on Build GRCh38
rs770465014 Chr.20: 45805067-45805067 on Build GRCh38
rs376281944 Chr.20: 45805091-45805091 on Build GRCh38
rs769099968 Chr.20: 45805095-45805095 on Build GRCh38
rs897091457 Chr.20: 45805159-45805159 on Build GRCh38
rs371721880 Chr.20: 45805133-45805133 on Build GRCh38
rs750661923 Chr.20: 45805195-45805195 on Build GRCh38
rs762237738 Chr.20: 45805112-45805112 on Build GRCh38
rs955868346 Chr.20: 45805059-45805059 on Build GRCh38
rs994537623 Chr.20: 45805174-45805174 on Build GRCh38
rs963221835 Chr.20: 45805209-45805209 on Build GRCh38
rs377178414 Chr.20: 45805096-45805096 on Build GRCh38
rs760744526 Chr.20: 45805138-45805138 on Build GRCh38
rs767168083 Chr.20: 45805128-45805128 on Build GRCh38
rs191065150 Chr.20: 45805087-45805087 on Build GRCh38
rs925597139 Chr.20: 45805052-45805052 on Build GRCh38
rs757061807 Chr.20: 45805172-45805172 on Build GRCh38
rs764061737 Chr.20: 45805139-45805139 on Build GRCh38
rs373336114 Chr.20: 45805081-45805081 on Build GRCh38
rs149931759 Chr.20: 45805094-45805094 on Build GRCh38

View Full Product Details