Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 19633718-19633971 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MPHOSPH8
Entrez Gene ID
54737
Gene Name
M-phase phosphoprotein 8
Gene Aliases
HSMPP8, TWA3, mpp8
UniGene
Hs.269654
Gene Chromosome Location
Chr.13: 19633647-19673459 on Build GRCh38
Transcripts
NM_017520.3 XM_011535124.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3955429 c.120A>G p.A40A Chr.13: 19633868-19633868 on Build GRCh38
3467580 c.40C>T p.P14S Chr.13: 19633788-19633788 on Build GRCh38
6571800 c.52G>C p.A18P Chr.13: 19633800-19633800 on Build GRCh38
7227095 c.120A>C p.A40A Chr.13: 19633868-19633868 on Build GRCh38
7417267 c.115G>A p.D39N Chr.13: 19633863-19633863 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs989361219 Chr.13: 19633898-19633898 on Build GRCh38
rs763545081 Chr.13: 19633779-19633779 on Build GRCh38
rs1057089 Chr.13: 19633838-19633838 on Build GRCh38
rs566133288 Chr.13: 19633755-19633755 on Build GRCh38
rs112488302 Chr.13: 19633813-19633813 on Build GRCh38
rs756737583 Chr.13: 19633748-19633748 on Build GRCh38
rs768452659 Chr.13: 19633748-19633749 on Build GRCh38
rs749787960 Chr.13: 19633762-19633762 on Build GRCh38
rs767045700 Chr.13: 19633946-19633946 on Build GRCh38
rs886613219 Chr.13: 19633772-19633772 on Build GRCh38
rs899291580 Chr.13: 19633894-19633894 on Build GRCh38
rs1057088 Chr.13: 19633828-19633828 on Build GRCh38
rs748221226 Chr.13: 19633784-19633784 on Build GRCh38
rs779448685 Chr.13: 19633872-19633872 on Build GRCh38
rs371343470 Chr.13: 19633871-19633871 on Build GRCh38
rs777783196 Chr.13: 19633895-19633895 on Build GRCh38
rs113455539 Chr.13: 19633805-19633805 on Build GRCh38
rs752612246 Chr.13: 19633911-19633911 on Build GRCh38
rs554482845 Chr.13: 19633947-19633947 on Build GRCh38
rs865869170 Chr.13: 19633782-19633782 on Build GRCh38
rs747253449 Chr.13: 19633799-19633799 on Build GRCh38
rs760187859 Chr.13: 19633949-19633949 on Build GRCh38
rs750073057 Chr.13: 19633837-19633837 on Build GRCh38
rs768611163 Chr.13: 19633925-19633925 on Build GRCh38
rs780856708 Chr.13: 19633749-19633749 on Build GRCh38
rs967712386 Chr.13: 19633791-19633791 on Build GRCh38
rs764072513 Chr.13: 19633809-19633809 on Build GRCh38
rs751021593 Chr.13: 19633743-19633743 on Build GRCh38
rs956456218 Chr.13: 19633738-19633738 on Build GRCh38
rs759629606 Chr.13: 19633909-19633909 on Build GRCh38
rs149968142 Chr.13: 19633937-19633937 on Build GRCh38
rs762856981 Chr.13: 19633808-19633808 on Build GRCh38
rs749050176 Chr.13: 19633922-19633922 on Build GRCh38
rs766021897 Chr.13: 19633848-19633848 on Build GRCh38
rs758491809 Chr.13: 19633879-19633879 on Build GRCh38
rs1050934661 Chr.13: 19633839-19633839 on Build GRCh38
rs199797432 Chr.13: 19633737-19633737 on Build GRCh38
rs748655472 Chr.13: 19633873-19633873 on Build GRCh38
rs777013902 Chr.13: 19633918-19633918 on Build GRCh38
rs776374764 Chr.13: 19633830-19633832 on Build GRCh38
rs755407839 Chr.13: 19633763-19633763 on Build GRCh38
rs773464705 Chr.13: 19633795-19633795 on Build GRCh38
rs139734802 Chr.13: 19633821-19633821 on Build GRCh38
rs774185324 Chr.13: 19633817-19633817 on Build GRCh38
rs761641080 Chr.13: 19633912-19633913 on Build GRCh38
rs571555886 Chr.13: 19633913-19633913 on Build GRCh38
rs774321633 Chr.13: 19633934-19633934 on Build GRCh38
rs905433183 Chr.13: 19633936-19633936 on Build GRCh38
rs146735121 Chr.13: 19633783-19633783 on Build GRCh38
rs775360707 Chr.13: 19633807-19633807 on Build GRCh38
rs755828085 Chr.13: 19633840-19633840 on Build GRCh38
rs548773739 Chr.13: 19633754-19633754 on Build GRCh38
rs772089186 Chr.13: 19633788-19633788 on Build GRCh38
rs201058071 Chr.13: 19633910-19633910 on Build GRCh38
rs1036414214 Chr.13: 19633887-19633887 on Build GRCh38
rs908383753 Chr.13: 19633853-19633853 on Build GRCh38
rs766984872 Chr.13: 19633836-19633836 on Build GRCh38

View Full Product Details