Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 80273652-80273925 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MAT1A
Entrez Gene ID
4143
Gene Name
methionine adenosyltransferase 1A
Gene Aliases
MAT, MATA1, SAMS, SAMS1
UniGene
Hs.282670
Gene Chromosome Location
Chr.10: 80271820-80290003 on Build GRCh38
Transcripts
NM_000429.2 XM_005269842.4 XM_005269843.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1297462 c.1160G>C p.W387S Chr.10: 80273809-80273809 on Build GRCh38
6309316 c.1119G>A p.K373K Chr.10: 80273850-80273850 on Build GRCh38
3440975 c.1090T>G p.L364V Chr.10: 80273879-80273879 on Build GRCh38
342761 c.1165G>C p.V389L Chr.10: 80273804-80273804 on Build GRCh38
5534319 c.1134C>T p.G378G Chr.10: 80273835-80273835 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs776948993 Chr.10: 80273808-80273808 on Build GRCh38
rs765379709 Chr.10: 80273817-80273817 on Build GRCh38
rs566176857 Chr.10: 80273815-80273815 on Build GRCh38
rs944215085 Chr.10: 80273862-80273862 on Build GRCh38
rs745711459 Chr.10: 80273803-80273803 on Build GRCh38
rs2993763 Chr.10: 80273838-80273838 on Build GRCh38
rs769060199 Chr.10: 80273797-80273797 on Build GRCh38
rs979162676 Chr.10: 80273680-80273680 on Build GRCh38
rs1032355447 Chr.10: 80273695-80273695 on Build GRCh38
rs886047319 Chr.10: 80273697-80273697 on Build GRCh38
rs770937613 Chr.10: 80273779-80273779 on Build GRCh38
rs747040359 Chr.10: 80273787-80273787 on Build GRCh38
rs200663190 Chr.10: 80273821-80273821 on Build GRCh38
rs752428447 Chr.10: 80273877-80273877 on Build GRCh38
rs960163320 Chr.10: 80273724-80273724 on Build GRCh38
rs773105969 Chr.10: 80273819-80273819 on Build GRCh38
rs987081828 Chr.10: 80273698-80273698 on Build GRCh38
rs755385387 Chr.10: 80273841-80273841 on Build GRCh38
rs757114301 Chr.10: 80273699-80273699 on Build GRCh38
rs201896138 Chr.10: 80273765-80273765 on Build GRCh38
rs7087728 Chr.10: 80273714-80273714 on Build GRCh38
rs373532399 Chr.10: 80273805-80273805 on Build GRCh38
rs78036669 Chr.10: 80273748-80273748 on Build GRCh38
rs376771993 Chr.10: 80273730-80273730 on Build GRCh38
rs957066588 Chr.10: 80273686-80273686 on Build GRCh38
rs375448565 Chr.10: 80273894-80273894 on Build GRCh38
rs774297257 Chr.10: 80273781-80273781 on Build GRCh38
rs116371248 Chr.10: 80273708-80273708 on Build GRCh38
rs892021803 Chr.10: 80273697-80273697 on Build GRCh38
rs777184287 Chr.10: 80273820-80273820 on Build GRCh38
rs758594127 Chr.10: 80273743-80273743 on Build GRCh38
rs200191278 Chr.10: 80273902-80273902 on Build GRCh38
rs781630967 Chr.10: 80273776-80273776 on Build GRCh38
rs199546548 Chr.10: 80273762-80273762 on Build GRCh38
rs980270209 Chr.10: 80273713-80273713 on Build GRCh38
rs368336046 Chr.10: 80273829-80273829 on Build GRCh38
rs748683813 Chr.10: 80273903-80273903 on Build GRCh38
rs1021299615 Chr.10: 80273708-80273708 on Build GRCh38
rs756826993 Chr.10: 80273904-80273904 on Build GRCh38
rs755691897 Chr.10: 80273763-80273763 on Build GRCh38
rs755332006 Chr.10: 80273738-80273738 on Build GRCh38
rs780881453 Chr.10: 80273754-80273754 on Build GRCh38
rs907813543 Chr.10: 80273742-80273742 on Build GRCh38
rs370084055 Chr.10: 80273769-80273769 on Build GRCh38
rs767838324 Chr.10: 80273871-80273871 on Build GRCh38
rs1050271601 Chr.10: 80273840-80273840 on Build GRCh38
rs1065766 Chr.10: 80273682-80273682 on Build GRCh38

View Full Product Details