Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 108377123-108377372 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HHLA2
Entrez Gene ID
11148
Gene Name
HERV-H LTR-associating 2
Gene Aliases
B7-H5, B7-H7, B7H7, B7y
UniGene
Hs.225968
Gene Chromosome Location
Chr.3: 108296490-108378285 on Build GRCh38
Transcripts
NM_001282556.1 NM_001282557.1 NM_001282558.1 NM_001282559.1 NM_007072.3 XM_005247080.3 XM_011512362.1 XM_011512363.1 XM_011512364.1 XM_011512367.2 XM_017005642.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6223535 c.1225-2A>G p.? Chr.3: 108377256-108377256 on Build GRCh38
6232494 c.1225-7C>G p.? Chr.3: 108377251-108377251 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs765883154 Chr.3: 108377191-108377191 on Build GRCh38
rs372303211 Chr.3: 108377214-108377214 on Build GRCh38
rs759215285 Chr.3: 108377230-108377230 on Build GRCh38
rs557671189 Chr.3: 108377196-108377196 on Build GRCh38
rs147822650 Chr.3: 108377228-108377228 on Build GRCh38
rs774556856 Chr.3: 108377225-108377225 on Build GRCh38
rs757917288 Chr.3: 108377290-108377290 on Build GRCh38
rs939368286 Chr.3: 108377297-108377297 on Build GRCh38
rs913734622 Chr.3: 108377152-108377152 on Build GRCh38
rs559616330 Chr.3: 108377247-108377247 on Build GRCh38
rs372264540 Chr.3: 108377258-108377258 on Build GRCh38
rs545651330 Chr.3: 108377243-108377243 on Build GRCh38
rs779464186 Chr.3: 108377269-108377269 on Build GRCh38
rs754205641 Chr.3: 108377253-108377253 on Build GRCh38
rs146238562 Chr.3: 108377165-108377165 on Build GRCh38
rs541700038 Chr.3: 108377303-108377303 on Build GRCh38
rs945487366 Chr.3: 108377160-108377160 on Build GRCh38
rs753575716 Chr.3: 108377205-108377205 on Build GRCh38
rs747591855 Chr.3: 108377288-108377288 on Build GRCh38
rs770527809 Chr.3: 108377299-108377299 on Build GRCh38
rs746704480 Chr.3: 108377298-108377298 on Build GRCh38
rs1043774628 Chr.3: 108377161-108377161 on Build GRCh38
rs368334820 Chr.3: 108377203-108377203 on Build GRCh38
rs992196678 Chr.3: 108377352-108377352 on Build GRCh38
rs1463426 Chr.3: 108377251-108377251 on Build GRCh38
rs777242133 Chr.3: 108377294-108377294 on Build GRCh38

View Full Product Details