Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 39856240-39856511 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DAAM2
Entrez Gene ID
23500
Gene Name
dishevelled associated activator of morphogenesis 2
Gene Aliases
dJ90A20A.1
UniGene
Hs.357128
Gene Chromosome Location
Chr.6: 39792366-39904877 on Build GRCh38
Transcripts
NM_001201427.1 NM_015345.3 XM_006715039.3 XM_006715040.3 XM_006715042.2 XM_006715043.2 XM_006715045.3 XM_006715046.3 XM_017010630.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5553370 c.109G>C p.E37Q Chr.6: 39856411-39856411 on Build GRCh38
6495078 c.9C>T p.P3P Chr.6: 39856311-39856311 on Build GRCh38
7363598 c.122C>G p.P41R Chr.6: 39856424-39856424 on Build GRCh38
1568365 c.11G>A p.R4H Chr.6: 39856313-39856313 on Build GRCh38
6547224 c.99G>T p.L33L Chr.6: 39856401-39856401 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs775379327 Chr.6: 39856313-39856313 on Build GRCh38
rs749092391 Chr.6: 39856393-39856393 on Build GRCh38
rs372308288 Chr.6: 39856480-39856480 on Build GRCh38
rs377508623 Chr.6: 39856428-39856428 on Build GRCh38
rs201680252 Chr.6: 39856325-39856325 on Build GRCh38
rs185320536 Chr.6: 39856311-39856311 on Build GRCh38
rs375245894 Chr.6: 39856435-39856435 on Build GRCh38
rs199767843 Chr.6: 39856354-39856354 on Build GRCh38
rs778290774 Chr.6: 39856286-39856286 on Build GRCh38
rs778326747 Chr.6: 39856356-39856356 on Build GRCh38
rs527543903 Chr.6: 39856322-39856322 on Build GRCh38
rs6936815 Chr.6: 39856484-39856484 on Build GRCh38
rs749464558 Chr.6: 39856454-39856454 on Build GRCh38
rs372928269 Chr.6: 39856371-39856371 on Build GRCh38
rs370984960 Chr.6: 39856431-39856431 on Build GRCh38
rs781749211 Chr.6: 39856443-39856443 on Build GRCh38
rs979601420 Chr.6: 39856366-39856366 on Build GRCh38
rs772573007 Chr.6: 39856430-39856430 on Build GRCh38
rs555296179 Chr.6: 39856449-39856449 on Build GRCh38
rs189802189 Chr.6: 39856312-39856312 on Build GRCh38
rs368227414 Chr.6: 39856436-39856436 on Build GRCh38
rs575112318 Chr.6: 39856474-39856474 on Build GRCh38
rs762893399 Chr.6: 39856320-39856320 on Build GRCh38
rs754638762 Chr.6: 39856288-39856288 on Build GRCh38
rs764968349 Chr.6: 39856306-39856307 on Build GRCh38
rs757660725 Chr.6: 39856439-39856439 on Build GRCh38
rs968166545 Chr.6: 39856363-39856363 on Build GRCh38
rs761588902 Chr.6: 39856416-39856416 on Build GRCh38
rs373643190 Chr.6: 39856349-39856349 on Build GRCh38
rs761373847 Chr.6: 39856326-39856326 on Build GRCh38
rs747105413 Chr.6: 39856296-39856296 on Build GRCh38
rs751949763 Chr.6: 39856358-39856358 on Build GRCh38
rs375928580 Chr.6: 39856372-39856372 on Build GRCh38
rs756332066 Chr.6: 39856452-39856452 on Build GRCh38
rs535365178 Chr.6: 39856434-39856434 on Build GRCh38
rs376650953 Chr.6: 39856353-39856353 on Build GRCh38
rs567170970 Chr.6: 39856342-39856342 on Build GRCh38
rs751981397 Chr.6: 39856438-39856438 on Build GRCh38
rs779209040 Chr.6: 39856265-39856265 on Build GRCh38
rs768631427 Chr.6: 39856395-39856395 on Build GRCh38
rs34924764 Chr.6: 39856388-39856388 on Build GRCh38
rs548086802 Chr.6: 39856309-39856309 on Build GRCh38
rs941887009 Chr.6: 39856302-39856302 on Build GRCh38
rs368905308 Chr.6: 39856281-39856281 on Build GRCh38
rs774209641 Chr.6: 39856396-39856396 on Build GRCh38
rs770971141 Chr.6: 39856307-39856307 on Build GRCh38
rs369889524 Chr.6: 39856347-39856347 on Build GRCh38
rs200592505 Chr.6: 39856385-39856385 on Build GRCh38
rs780412197 Chr.6: 39856453-39856453 on Build GRCh38
rs11965120 Chr.6: 39856329-39856329 on Build GRCh38
rs745986379 Chr.6: 39856384-39856384 on Build GRCh38
rs758879407 Chr.6: 39856282-39856282 on Build GRCh38

View Full Product Details