Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 98914704-98914977 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PLPPR5
Entrez Gene ID
163404
Gene Name
phospholipid phosphatase related 5
Gene Aliases
LPPR5, PAP2, PAP2D, PRG5
UniGene
Hs.483948
Gene Chromosome Location
Chr.1: 98890245-99005850 on Build GRCh38
Transcripts
NM_001010861.2 NM_001037317.1 XM_011540836.2 XM_011540837.1 XM_011540838.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3786294 c.933G>T p.Q311H Chr.1: 98914786-98914786 on Build GRCh38
5847549 c.913G>A p.E305K Chr.1: 98914806-98914806 on Build GRCh38
4964915 c.869C>T p.A290V Chr.1: 98914850-98914850 on Build GRCh38
3769046 c.873G>A p.Q291Q Chr.1: 98914846-98914846 on Build GRCh38
2261490 c.808G>T p.V270L Chr.1: 98914911-98914911 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs759445624 Chr.1: 98914812-98914812 on Build GRCh38
rs776103466 Chr.1: 98914884-98914884 on Build GRCh38
rs758813172 Chr.1: 98914857-98914857 on Build GRCh38
rs765261785 Chr.1: 98914760-98914761 on Build GRCh38
rs201725625 Chr.1: 98914823-98914823 on Build GRCh38
rs879811409 Chr.1: 98914869-98914869 on Build GRCh38
rs749786089 Chr.1: 98914738-98914738 on Build GRCh38
rs766549502 Chr.1: 98914830-98914830 on Build GRCh38
rs143279943 Chr.1: 98914838-98914838 on Build GRCh38
rs748613706 Chr.1: 98914755-98914755 on Build GRCh38
rs770457865 Chr.1: 98914794-98914794 on Build GRCh38
rs779410458 Chr.1: 98914749-98914749 on Build GRCh38
rs755540648 Chr.1: 98914834-98914834 on Build GRCh38
rs375842420 Chr.1: 98914766-98914766 on Build GRCh38
rs199656479 Chr.1: 98914854-98914854 on Build GRCh38
rs373204041 Chr.1: 98914822-98914822 on Build GRCh38
rs112548482 Chr.1: 98914781-98914781 on Build GRCh38
rs750591473 Chr.1: 98914792-98914795 on Build GRCh38
rs753904311 Chr.1: 98914930-98914930 on Build GRCh38
rs766502806 Chr.1: 98914952-98914953 on Build GRCh38
rs756448637 Chr.1: 98914824-98914824 on Build GRCh38
rs969473674 Chr.1: 98914868-98914868 on Build GRCh38
rs763140255 Chr.1: 98914942-98914943 on Build GRCh38
rs770415284 Chr.1: 98914758-98914758 on Build GRCh38
rs777252432 Chr.1: 98914860-98914860 on Build GRCh38
rs529772368 Chr.1: 98914832-98914832 on Build GRCh38
rs754032121 Chr.1: 98914833-98914833 on Build GRCh38
rs139357152 Chr.1: 98914814-98914814 on Build GRCh38
rs774066694 Chr.1: 98914912-98914912 on Build GRCh38
rs759703522 Chr.1: 98914937-98914937 on Build GRCh38
rs769650634 Chr.1: 98914889-98914889 on Build GRCh38
rs62640872 Chr.1: 98914891-98914891 on Build GRCh38
rs746641121 Chr.1: 98914862-98914862 on Build GRCh38
rs765615809 Chr.1: 98914943-98914943 on Build GRCh38
rs996331901 Chr.1: 98914734-98914734 on Build GRCh38
rs566350611 Chr.1: 98914817-98914817 on Build GRCh38
rs760969705 Chr.1: 98914918-98914918 on Build GRCh38
rs776158487 Chr.1: 98914798-98914798 on Build GRCh38
rs780655287 Chr.1: 98914736-98914736 on Build GRCh38
rs763977677 Chr.1: 98914911-98914911 on Build GRCh38
rs535457594 Chr.1: 98914779-98914779 on Build GRCh38
rs1013573601 Chr.1: 98914795-98914795 on Build GRCh38
rs745448777 Chr.1: 98914888-98914888 on Build GRCh38
rs755594206 Chr.1: 98914743-98914743 on Build GRCh38
rs770619194 Chr.1: 98914871-98914871 on Build GRCh38
rs766778481 Chr.1: 98914919-98914919 on Build GRCh38
rs761883609 Chr.1: 98914753-98914756 on Build GRCh38

View Full Product Details