Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 15445530-15445787 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CTRC
Entrez Gene ID
11330
Gene Name
chymotrypsin C
Gene Aliases
CLCR, ELA4
UniGene
Hs.631869
Gene Chromosome Location
Chr.1: 15438442-15449247 on Build GRCh38
Transcripts
NM_007272.2 XM_011540550.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7260267 c.792+10G>A p.? Chr.1: 15445759-15445759 on Build GRCh38
5377998 c.643G>A p.D215N Chr.1: 15445600-15445600 on Build GRCh38
5483604 c.649G>A p.G217S Chr.1: 15445606-15445606 on Build GRCh38
3475817 c.684C>T p.S228S Chr.1: 15445641-15445641 on Build GRCh38
3475816 c.680G>A p.G227D Chr.1: 15445637-15445637 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs763435999 Chr.1: 15445551-15445551 on Build GRCh38
rs1013133526 Chr.1: 15445598-15445598 on Build GRCh38
rs1005522576 Chr.1: 15445697-15445697 on Build GRCh38
rs200092422 Chr.1: 15445558-15445558 on Build GRCh38
rs764039167 Chr.1: 15445748-15445748 on Build GRCh38
rs758245744 Chr.1: 15445559-15445559 on Build GRCh38
rs199897030 Chr.1: 15445555-15445555 on Build GRCh38
rs777187660 Chr.1: 15445737-15445737 on Build GRCh38
rs147925927 Chr.1: 15445694-15445694 on Build GRCh38
rs567745213 Chr.1: 15445636-15445636 on Build GRCh38
rs571960316 Chr.1: 15445555-15445555 on Build GRCh38
rs759881879 Chr.1: 15445688-15445688 on Build GRCh38
rs772573958 Chr.1: 15445724-15445724 on Build GRCh38
rs142027137 Chr.1: 15445637-15445637 on Build GRCh38
rs202058123 Chr.1: 15445606-15445606 on Build GRCh38
rs121909293 Chr.1: 15445717-15445717 on Build GRCh38
rs191025876 Chr.1: 15445557-15445557 on Build GRCh38
rs371799801 Chr.1: 15445761-15445761 on Build GRCh38
rs754538818 Chr.1: 15445553-15445554 on Build GRCh38
rs202049497 Chr.1: 15445583-15445583 on Build GRCh38
rs756384572 Chr.1: 15445743-15445744 on Build GRCh38
rs757757145 Chr.1: 15445562-15445563 on Build GRCh38
rs368477746 Chr.1: 15445584-15445584 on Build GRCh38
rs766713372 Chr.1: 15445591-15445591 on Build GRCh38
rs111372278 Chr.1: 15445720-15445720 on Build GRCh38
rs515726210 Chr.1: 15445695-15445718 on Build GRCh38
rs200406696 Chr.1: 15445726-15445726 on Build GRCh38
rs183053579 Chr.1: 15445585-15445585 on Build GRCh38
rs760962602 Chr.1: 15445676-15445676 on Build GRCh38
rs543849938 Chr.1: 15445555-15445556 on Build GRCh38
rs753912502 Chr.1: 15445593-15445593 on Build GRCh38
rs777436696 Chr.1: 15445679-15445679 on Build GRCh38
rs751482739 Chr.1: 15445751-15445751 on Build GRCh38
rs771229796 Chr.1: 15445678-15445678 on Build GRCh38
rs367676653 Chr.1: 15445675-15445675 on Build GRCh38
rs777470722 Chr.1: 15445560-15445560 on Build GRCh38
rs772523751 Chr.1: 15445661-15445661 on Build GRCh38
rs757062706 Chr.1: 15445714-15445714 on Build GRCh38
rs10927786 Chr.1: 15445562-15445562 on Build GRCh38
rs780832496 Chr.1: 15445640-15445640 on Build GRCh38
rs781447617 Chr.1: 15445679-15445679 on Build GRCh38
rs769975164 Chr.1: 15445745-15445745 on Build GRCh38
rs769089707 Chr.1: 15445575-15445575 on Build GRCh38
rs780783336 Chr.1: 15445715-15445715 on Build GRCh38
rs140993290 Chr.1: 15445660-15445660 on Build GRCh38
rs748753747 Chr.1: 15445719-15445719 on Build GRCh38
rs779469584 Chr.1: 15445659-15445659 on Build GRCh38
rs200412314 Chr.1: 15445693-15445693 on Build GRCh38
rs759821387 Chr.1: 15445740-15445740 on Build GRCh38
rs373970317 Chr.1: 15445554-15445555 on Build GRCh38
rs755811899 Chr.1: 15445718-15445718 on Build GRCh38
rs769324715 Chr.1: 15445656-15445656 on Build GRCh38
rs201486613 Chr.1: 15445631-15445631 on Build GRCh38
rs186735838 Chr.1: 15445556-15445556 on Build GRCh38
rs777608312 Chr.1: 15445635-15445635 on Build GRCh38
rs755130943 Chr.1: 15445605-15445605 on Build GRCh38
rs113257816 Chr.1: 15445712-15445712 on Build GRCh38
rs762952174 Chr.1: 15445746-15445746 on Build GRCh38
rs947267758 Chr.1: 15445734-15445734 on Build GRCh38
rs745383368 Chr.1: 15445568-15445568 on Build GRCh38
rs778166573 Chr.1: 15445725-15445725 on Build GRCh38
rs540753875 Chr.1: 15445735-15445735 on Build GRCh38
rs752651686 Chr.1: 15445626-15445626 on Build GRCh38
rs75971387 Chr.1: 15445647-15445647 on Build GRCh38
rs142560329 Chr.1: 15445703-15445703 on Build GRCh38
rs150078209 Chr.1: 15445704-15445704 on Build GRCh38

View Full Product Details