Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 102367864-102368105 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IL18R1
Entrez Gene ID
8809
Gene Name
interleukin 18 receptor 1
Gene Aliases
CD218a, CDw218a, IL-1Rrp, IL18RA, IL1RRP
UniGene
Hs.469521
Gene Chromosome Location
Chr.2: 102355817-102398777 on Build GRCh38
Transcripts
NM_001282399.1 NM_003855.3 XM_005264039.3 XM_005264040.3 XM_011512099.1 XM_017005181.1 XM_017005182.1 XM_017005183.1 XM_017005184.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
257160 c.164G>A p.S55N Chr.2: 102367930-102367930 on Build GRCh38
240219 c.185C>T p.S62L Chr.2: 102367951-102367951 on Build GRCh38
7422028 c.202C>A p.L68M Chr.2: 102367968-102367968 on Build GRCh38
6152975 c.166T>G p.W56G Chr.2: 102367932-102367932 on Build GRCh38
219971 c.238G>T p.D80Y Chr.2: 102368004-102368004 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs375398936 Chr.2: 102367904-102367904 on Build GRCh38
rs181278302 Chr.2: 102368018-102368018 on Build GRCh38
rs766098022 Chr.2: 102367974-102367974 on Build GRCh38
rs752359007 Chr.2: 102367947-102367947 on Build GRCh38
rs56183212 Chr.2: 102367892-102367892 on Build GRCh38
rs917316315 Chr.2: 102367891-102367891 on Build GRCh38
rs777908870 Chr.2: 102367925-102367925 on Build GRCh38
rs200299555 Chr.2: 102368065-102368065 on Build GRCh38
rs368716787 Chr.2: 102367919-102367919 on Build GRCh38
rs765900628 Chr.2: 102368003-102368003 on Build GRCh38
rs779351136 Chr.2: 102367917-102367917 on Build GRCh38
rs778234725 Chr.2: 102368039-102368039 on Build GRCh38
rs951497108 Chr.2: 102367894-102367894 on Build GRCh38
rs781182846 Chr.2: 102368063-102368063 on Build GRCh38
rs909864674 Chr.2: 102367931-102367931 on Build GRCh38
rs1033414496 Chr.2: 102367973-102367973 on Build GRCh38
rs761549416 Chr.2: 102367988-102367988 on Build GRCh38
rs763770123 Chr.2: 102367978-102367978 on Build GRCh38
rs758012132 Chr.2: 102367987-102367987 on Build GRCh38
rs139816270 Chr.2: 102367960-102367960 on Build GRCh38
rs757515515 Chr.2: 102368055-102368055 on Build GRCh38
rs1039891306 Chr.2: 102367976-102367976 on Build GRCh38
rs766876534 Chr.2: 102367995-102367995 on Build GRCh38
rs535384643 Chr.2: 102367924-102367924 on Build GRCh38
rs367800541 Chr.2: 102367968-102367968 on Build GRCh38
rs749953966 Chr.2: 102368000-102368000 on Build GRCh38
rs770879570 Chr.2: 102367951-102367951 on Build GRCh38
rs773834061 Chr.2: 102367981-102367981 on Build GRCh38
rs760113124 Chr.2: 102368002-102368002 on Build GRCh38
rs776795173 Chr.2: 102367956-102367956 on Build GRCh38
rs751687643 Chr.2: 102368050-102368050 on Build GRCh38
rs370305765 Chr.2: 102367955-102367955 on Build GRCh38

View Full Product Details