Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 98104430-98104695 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RIDA
Entrez Gene ID
10247
Gene Name
reactive intermediate imine deaminase A homolog
Gene Aliases
HRSP12, P14.5, PSP, UK114
UniGene
Hs.18426
Gene Chromosome Location
Chr.8: 98102339-98117190 on Build GRCh38
Transcripts
NM_005836.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
69253 c.296-1G>C p.? Chr.8: 98104545-98104545 on Build GRCh38
5875176 c.313C>T p.P105S Chr.8: 98104527-98104527 on Build GRCh38
1643672 c.351A>C p.K117N Chr.8: 98104489-98104489 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs763397255 Chr.8: 98104571-98104571 on Build GRCh38
rs780684980 Chr.8: 98104564-98104565 on Build GRCh38
rs367867762 Chr.8: 98104610-98104610 on Build GRCh38
rs762265538 Chr.8: 98104480-98104480 on Build GRCh38
rs759160435 Chr.8: 98104523-98104523 on Build GRCh38
rs925569936 Chr.8: 98104554-98104554 on Build GRCh38
rs759082130 Chr.8: 98104464-98104464 on Build GRCh38
rs770776366 Chr.8: 98104517-98104517 on Build GRCh38
rs781173197 Chr.8: 98104501-98104501 on Build GRCh38
rs765774426 Chr.8: 98104556-98104556 on Build GRCh38
rs368858904 Chr.8: 98104536-98104536 on Build GRCh38
rs751037800 Chr.8: 98104566-98104566 on Build GRCh38
rs754345999 Chr.8: 98104479-98104479 on Build GRCh38
rs769565768 Chr.8: 98104510-98104510 on Build GRCh38
rs780420240 Chr.8: 98104490-98104490 on Build GRCh38
rs977177686 Chr.8: 98104612-98104612 on Build GRCh38
rs760247414 Chr.8: 98104547-98104547 on Build GRCh38
rs777699034 Chr.8: 98104515-98104515 on Build GRCh38
rs373486688 Chr.8: 98104537-98104537 on Build GRCh38
rs749012373 Chr.8: 98104516-98104516 on Build GRCh38
rs987302040 Chr.8: 98104466-98104466 on Build GRCh38
rs146537203 Chr.8: 98104499-98104499 on Build GRCh38
rs949552165 Chr.8: 98104628-98104628 on Build GRCh38
rs935580616 Chr.8: 98104607-98104607 on Build GRCh38
rs758839013 Chr.8: 98104484-98104484 on Build GRCh38
rs773993780 Chr.8: 98104462-98104462 on Build GRCh38
rs766902349 Chr.8: 98104581-98104581 on Build GRCh38
rs964391859 Chr.8: 98104509-98104509 on Build GRCh38
rs760050113 Chr.8: 98104583-98104583 on Build GRCh38
rs755777779 Chr.8: 98104589-98104593 on Build GRCh38
rs750761252 Chr.8: 98104481-98104481 on Build GRCh38
rs766993350 Chr.8: 98104470-98104470 on Build GRCh38
rs745434901 Chr.8: 98104575-98104579 on Build GRCh38
rs751892114 Chr.8: 98104491-98104491 on Build GRCh38
rs991172389 Chr.8: 98104533-98104533 on Build GRCh38
rs773978185 Chr.8: 98104520-98104520 on Build GRCh38
rs748294583 Chr.8: 98104502-98104502 on Build GRCh38

View Full Product Details