Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 48997024-48997249 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DDN
Entrez Gene ID
23109
Gene Name
dendrin
Gene Aliases
-
UniGene
Hs.591044
Gene Chromosome Location
Chr.12: 48995149-49001772 on Build GRCh38
Transcripts
NM_015086.1 XM_011538055.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4401909 c.1551A>G p.Q517Q Chr.12: 48997163-48997163 on Build GRCh38
6553238 c.1751C>A p.A584D Chr.12: 48997125-48997125 on Build GRCh38
4470473 c.1662C>T p.P554P Chr.12: 48997214-48997214 on Build GRCh38
4397215 c.1663G>A p.E555K Chr.12: 48997213-48997213 on Build GRCh38
6641344 c.1524delC p.T509fs*11 Chr.12: 48997190-48997190 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs570866519 Chr.12: 48997053-48997053 on Build GRCh38
rs778119790 Chr.12: 48997214-48997214 on Build GRCh38
rs778848522 Chr.12: 48997166-48997166 on Build GRCh38
rs763166047 Chr.12: 48997103-48997103 on Build GRCh38
rs753561904 Chr.12: 48997083-48997083 on Build GRCh38
rs779527092 Chr.12: 48997079-48997079 on Build GRCh38
rs774753545 Chr.12: 48997115-48997115 on Build GRCh38
rs781301608 Chr.12: 48997093-48997093 on Build GRCh38
rs748085787 Chr.12: 48997090-48997090 on Build GRCh38
rs772522583 Chr.12: 48997231-48997231 on Build GRCh38
rs752580713 Chr.12: 48997168-48997168 on Build GRCh38
rs969163559 Chr.12: 48997052-48997052 on Build GRCh38
rs370893823 Chr.12: 48997087-48997087 on Build GRCh38
rs539447931 Chr.12: 48997054-48997054 on Build GRCh38
rs1044956305 Chr.12: 48997174-48997174 on Build GRCh38
rs758388425 Chr.12: 48997179-48997179 on Build GRCh38
rs780194328 Chr.12: 48997209-48997209 on Build GRCh38
rs748370429 Chr.12: 48997123-48997124 on Build GRCh38
rs374013585 Chr.12: 48997218-48997218 on Build GRCh38
rs749963657 Chr.12: 48997074-48997074 on Build GRCh38
rs762299549 Chr.12: 48997118-48997118 on Build GRCh38
rs772203032 Chr.12: 48997092-48997092 on Build GRCh38
rs769694264 Chr.12: 48997200-48997200 on Build GRCh38
rs749495543 Chr.12: 48997213-48997213 on Build GRCh38
rs910575228 Chr.12: 48997187-48997187 on Build GRCh38
rs755773559 Chr.12: 48997075-48997075 on Build GRCh38
rs757772276 Chr.12: 48997068-48997068 on Build GRCh38
rs777521845 Chr.12: 48997191-48997191 on Build GRCh38
rs768930397 Chr.12: 48997216-48997216 on Build GRCh38
rs149836945 Chr.12: 48997140-48997140 on Build GRCh38
rs745983761 Chr.12: 48997094-48997094 on Build GRCh38
rs202028952 Chr.12: 48997225-48997225 on Build GRCh38
rs940434537 Chr.12: 48997091-48997091 on Build GRCh38
rs754737222 Chr.12: 48997156-48997156 on Build GRCh38
rs752998382 Chr.12: 48997084-48997084 on Build GRCh38
rs767946140 Chr.12: 48997123-48997123 on Build GRCh38
rs769939532 Chr.12: 48997095-48997095 on Build GRCh38
rs746997363 Chr.12: 48997196-48997196 on Build GRCh38
rs760068418 Chr.12: 48997125-48997125 on Build GRCh38
rs768948699 Chr.12: 48997111-48997111 on Build GRCh38
rs775533762 Chr.12: 48997097-48997097 on Build GRCh38
rs750966504 Chr.12: 48997124-48997124 on Build GRCh38
rs905112329 Chr.12: 48997188-48997188 on Build GRCh38
rs11834471 Chr.12: 48997141-48997141 on Build GRCh38
rs542931185 Chr.12: 48997116-48997116 on Build GRCh38
rs139973689 Chr.12: 48997129-48997129 on Build GRCh38

View Full Product Details