Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 46929898-46930153 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CYP4A11
Entrez Gene ID
1579
Gene Name
cytochrome P450 family 4 subfamily A member 11
Gene Aliases
CP4Y, CYP4A2, CYP4AII
UniGene
Hs.1645
Gene Chromosome Location
Chr.1: 46929174-46941484 on Build GRCh38
Transcripts
NM_000778.3 NM_001319155.1 NR_134988.1 NR_134989.1 NR_134990.1 NR_134991.1 NR_134992.1 NR_134993.1 NR_134994.1 XM_017000464.1 XM_017000465.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7187051 c.1550A>G p.D517G Chr.1: 46930125-46930125 on Build GRCh38
6883057 c.1549G>A p.D517N Chr.1: 46930126-46930126 on Build GRCh38
7279061 c.1529C>A p.P510H Chr.1: 46930146-46930146 on Build GRCh38
3490394 c.1529C>T p.P510L Chr.1: 46930146-46930146 on Build GRCh38
4165226 c.1554G>T p.Q518H Chr.1: 46930121-46930121 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs750003772 Chr.1: 46930080-46930080 on Build GRCh38
rs770090187 Chr.1: 46930069-46930069 on Build GRCh38
rs760231988 Chr.1: 46930084-46930084 on Build GRCh38
rs535455348 Chr.1: 46929965-46929965 on Build GRCh38
rs916427561 Chr.1: 46929975-46929976 on Build GRCh38
rs766104878 Chr.1: 46930086-46930086 on Build GRCh38
rs371480688 Chr.1: 46930099-46930099 on Build GRCh38
rs114160471 Chr.1: 46930046-46930046 on Build GRCh38
rs763466642 Chr.1: 46930073-46930073 on Build GRCh38
rs775929602 Chr.1: 46930071-46930071 on Build GRCh38
rs1034944219 Chr.1: 46929925-46929925 on Build GRCh38
rs990666488 Chr.1: 46929963-46929963 on Build GRCh38
rs59465754 Chr.1: 46929945-46929945 on Build GRCh38
rs912209394 Chr.1: 46930066-46930066 on Build GRCh38
rs371275764 Chr.1: 46930113-46930113 on Build GRCh38
rs918337365 Chr.1: 46930019-46930019 on Build GRCh38
rs772737025 Chr.1: 46930080-46930080 on Build GRCh38
rs974271490 Chr.1: 46930011-46930011 on Build GRCh38
rs747166920 Chr.1: 46930120-46930120 on Build GRCh38
rs944953368 Chr.1: 46930070-46930070 on Build GRCh38
rs753544364 Chr.1: 46930094-46930094 on Build GRCh38
rs370669636 Chr.1: 46930051-46930051 on Build GRCh38
rs9333037 Chr.1: 46930054-46930054 on Build GRCh38
rs962554653 Chr.1: 46930005-46930005 on Build GRCh38
rs900693133 Chr.1: 46930076-46930076 on Build GRCh38
rs758411083 Chr.1: 46930106-46930106 on Build GRCh38
rs752621683 Chr.1: 46930104-46930104 on Build GRCh38
rs946896473 Chr.1: 46930077-46930077 on Build GRCh38

View Full Product Details